Skip to Content
Merck
All Photos(2)

Key Documents

MABE163

Sigma-Aldrich

Anti-ASF-1 (SF2) Antibody, clone AK96

clone AK96, from mouse

Synonym(s):

splicing factor, arginine/serine-rich 1, Alternative-splicing factor 1, pre-mRNA-splicing factor SF2, P33 subunit, splicing factor 22, ASF-1, SF2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$567.00

$567.00


Estimated to ship on04 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$567.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$567.00


Estimated to ship on04 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AK96, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (immunogen homology), mouse (immunogen homology)

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SRSF1(6426)

General description

Alternative-splicing factor 1 (ASF-1), also known as SRp30a or SF2, is a member of the highly conserved family of SR proteins, a group of serine/arginine-rich domain-containing proteins that play a critical role in regulated and constitutive splicing of precursor mRNA. The SR family of proteins is characterized as having one to two RNA recognition motifs located in the n-terminal sequence, and an RS domain within the C-terminus that is rich in serine/arginine dipeptide repeats. ASF-1 has an inhibitory role in exon skipping, in order to maintain splicing accuracy and to monitor alternative splicing. Studies have shown that ASF-1 plays a role in the regulation of caspase-9 alternative splicing as well as being a critical component for ceramide to function as a mediator of alternative splicing for the same caspase.

Specificity

This antibody recognizes the N-terminus of ASF-1 (SF2).

Immunogen

Epitope: N-terminus
MBP-tagged recombinant protein corresponding to the N-terminus of human ASF-1 (SF2).

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected ASF-1 (SF2) in human prostate carcinoma cells and human testis tissue.
Western Blotting Analysis: A representative lot was used by an independent laboratory in western blotting (Sinha, R., et al. (2010). Mol. Cell. Biol. doi:10.1128/MCB.01270-09).
Immunoprecipitation Analysis: A representative lot immunoprecipitated ASF-1 (SF2) in HeLa nuclear extracts (Jablonski, J. A., et al. (2010). Nucleic Acids Research. 38(4):1249-1260).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins
Use Anti-ASF-1 (SF2) Antibody, clone AK96 (mouse monoclonal antibody) validated in WB, IP, ICC, IHC to detect ASF-1 (SF2) also known as splicing factor arginine/serine-rich 1, Alternative-splicing factor 1 & pre-mRNA-splicing factor SF2 P33 subunit

Quality

Evaluated by Western Blot in human lymph node lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected ASF-1 (SF2) on 10 µg of human lymph node lysate.

Target description

~ 30 kDa observed

Linkage

Replaces: MABE512

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl without 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human lymph node lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ningyang Yuan et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 11(21), e2306871-e2306871 (2024-04-04)
RNA splicing dysregulation and the involvement of specific splicing factors are emerging as common factors in both obesity and metabolic disorders. The study provides compelling evidence that the absence of the splicing factor SRSF1 in mature adipocytes results in whitening
Thomas C Whisenant et al.
PloS one, 10(12), e0144409-e0144409 (2015-12-08)
Activation of CD4 T cells is a reaction to challenges such as microbial pathogens, cancer and toxins that defines adaptive immune responses. The roles of T cell receptor crosslinking, intracellular signaling, and transcription factor activation are well described, but the
Soniya Dhanjal et al.
The Journal of biological chemistry, 290(21), 13354-13371 (2015-04-17)
In order to identify cellular factors that regulate human papillomavirus type 16 (HPV16) gene expression, cervical cancer cells permissive for HPV16 late gene expression were identified and characterized. These cells either contained a novel spliced variant of the L1 mRNAs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service