Skip to Content
Merck
All Photos(1)

Key Documents

MABE1152M

Sigma-Aldrich

Anti-Cryptochrome-1 Antibody, clone D46-3361

clone D46-3361, from rat

Synonym(s):

Cry1, cryptochrome 1 (photolyase-like), Cryptochrome-1, Phll1, Cry-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$214.00
100 μL
$512.00

$214.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$214.00
100 μL
$512.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Check Cart for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

D46-3361, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μL

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Cry1(12952)

General description

Cryptochrome-1 (UniProt: P97784) is encoded by the Cry1 gene (Gene ID: 12952) in murine species. Cryptochrome-1 is an evolutionarily conserved member of the flavoprotein superfamily that acts as a transcriptional repressor and forms a core component of the circadian clock. It has a photolyase domain at its N-terminal (aa 3-132). Cryptochrome-1 has three FAD biding sites (aa 252, 289, and 355). However, only a minority of the protein molecules contain bound FAD. Transcription and translation of core clock components (CLOCK, NPAS2, ARNTL/BMAL1, ARNTL2/BMAL2, PER1, PER2, PER3, CRY1 and CRY2) play a critical role in rhythm generation, whereas delays imposed by post-translational modifications (PTMs) are important for determining the period (tau) of the rhythms. Disruptions in the circadian rhythms contribute to the pathology of cardiovascular diseases, cancer, metabolic syndrome, and aging. Crytopchrome-1 translocates to the nucleus through interaction with other clock proteins such as PER2 or ARNTL/BMAL1. The core clock genes: PER1/2/3 and CRY1/2, which are transcriptional repressors form the negative limb of the feedback loop and interact with the CLOCK|NPAS2-ARNTL/BMAL1|ARNTL2/BMAL2 heterodimer inhibiting its activity and thereby negatively regulating their own expression. Transcription factors, CLOCK or NPAS2 and ARNTL/BMAL1 or ARNTL2/BMAL2, form the positive limb of the feedback loop and act in the form of a heterodimer and activate the transcription of core clock genes and clock-controlled genes. It has been suggested that cryptochrome may act as a molecular gatekeeper to maintain CLOCK-ARNTL/BMAL1 in a poised and repressed state until the proper time for transcriptional activation. Animals with defective cryptochrome-1 display disruption in their glucose homeostasis with elevated blood glucose in response to acute feeding after an overnight fast and severely impaired glucose clearance in a glucose tolerance test.

Specificity

Clone D46-3361 is a rat monoclonal antibody that specifically detects Cryptochrome-1 in murine species. it targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of murine Cryptochrome-1.

Application

Anti-Cryptochrome-1, clone D46-3361 Antibody, Cat. No. MABE1152, is a highly specific rat monoclonal antibody that targets Cryptochrome-1 and has been tested for use in Chromatin Immunoprecipitation (ChIP) and Western Blotting.
Western Blotting Analysis: A representative lot detected Cryptochrome-1 in Western Blotting applications (Partch, C.L., et. al. (2006). Proc Natl Acad Sci USA. 103(27):10467-72; Ye, R., et. al. (2011). J Biol Chem. 286(29):25891-902; Ozturk, N., et. al. (2009). Proc Natl Acad Sci USA. 106(8):2841-6; Chiou, Y.Y., et. al. (2016). Proc Natl Acad Sci USA. 113(41):E6072-E6079).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Cryptochrome-1 in Chromatin Immunoprecipitation applications (Ye, R., et. al. (2011). J Biol Chem. 286(29):25891-902).

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Cryptochrome-1 in 10 µg of NIH/3T3 cell lysate.

Target description

~68 kDa observed; 68.0 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Purified rat monoclonal antibody IgM in PBS with 0.05% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service