Skip to Content
Merck
All Photos(4)

Key Documents

PRS4887

Sigma-Aldrich

Anti-IL-17 (ab2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CTLA8, Anti-Interleukin 17A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$646.00

$646.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1708


Select a Size

Change View
100 μG
$646.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$646.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1708

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL17A(3605)

General description

IL-17A (interleukin 17A) is a proinflammatory cytokine of Th17 (helper T cells). It is located on chromosome 6p12.

Immunogen

a 19 amino acid peptide from near the center of human IL-17A.

Biochem/physiol Actions

IL-17A (interleukin 17A) plays a major role in the pathogenesis of autoimmune diseases like multiple sclerosis and rheumatoid arthritis. It also plays a pathogenic role in allergic rhinitis (AR). It helps in the formation of several chemokines and cytokines in airway epithelial cells.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4887.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-17A and Th17 cells in lung inflammation: an update on the role of Th17 cell differentiation and IL-17R signaling in host defense against infection
Tsai HC, et al.
Clinical & Developmental Immunology (2013)
Satsuki Shimizu et al.
Scientific reports, 13(1), 5041-5041 (2023-03-29)
Infantile skin problems not only cause temporary pain and discomfort, but also have a long-term impact on health. Hence, the purpose of this cross-sectional study was to clarify the relationship between inflammatory cytokines and Malassezia fungal facial skin problems in
Foxp3 inhibits RORgammat-mediated IL-17A mRNA transcription through direct interaction with RORgammat
Ichiyama K, et al.
The Journal of Biological Chemistry, 283(25), 17003-17008 (2008)
Association between polymorphisms in cytokine genes IL-17A and IL-17F and development of allergic rhinitis and comorbid asthma in Chinese subjects
Wang M, et al.
Human immunology, 73(6), 647-653 (2012)
Tianxi Wang et al.
Experimental and therapeutic medicine, 21(6), 654-654 (2021-05-11)
Gastric carcinoma is a common type of gastrointestinal tumor with high morbidity and mortality rates. IL-17 is a newly discovered cytokine that has been reported to serve an important role in the development of gastric carcinoma. The potential effect of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service