Skip to Content
Merck
All Photos(2)

Key Documents

MABE1020

Sigma-Aldrich

Anti-Acetyl Lysine, Pan, clone RM101 Antibody

clone RM101, from rabbit, purified by affinity chromatography

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$875.00

$875.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$875.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$875.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM101, monoclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

all (based on 100% sequence homology)

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

General description

DNA is tightly packed in the nucleus into nucleosomes, generating an environment which is highly repressive towards DNA processes such as transcription. Acetylation of lysine residues within proteins has emerged as an important mechanism used by cells to overcome this repression. The acetylation of non-histone proteins such as transcription factors, as well as histones appears to be involved in this process. Acetylation may result in structural transitions as well as specific signaling within discrete chromatin domains. The role of acetylation in intracellular signaling has been inferred from the binding of acetylated peptides by the conserved bromodomain. Furthermore, recent findings suggest that bromodomain/acetylated-lysine recognition can serve as a regulatory mechanism in protein-protein interactions in numerous cellular processes such as chromatin remodeling and transcriptional activation.

Immunogen

Acetyl lysine-BSA corresponding to all species.

Application

Anti-Acetyl Lysine, Pan, clone RM101 is an antibody against Acetyl Lysine, Pan for use in WB, Dot Blot, ICC, IP.
Immunocytochemistry Analysis: A representative lot detected Acetyl Lysine, Pan in A431 cells.
Dot Blot Analysis: A 1:2,000 dilution from a representative lot detected Acetyl Lysine, Pan in an Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and an Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).
Immunoprecipitation Analysis: A 1:500 dilution from a representative lot immunoprecipitated Acetyl Lysine, Pan in A431 cell lysate.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors

Quality

Evaluated by Western Blotting in Trichostatin A (TSA) treated HeLa cell lysate.

Western Blotting Analysis: A 1:2,000 of this antibody detected Acetyl Lysine, Pan in Trichostatin A (TSA) treated HeLa cell lysate.

Physical form

Protein A purified
Purified rabbit monoclonal IgG in buffer containing PBS with 1% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service