Skip to Content
Merck
All Photos(1)

Key Documents

CAX2-MXT20

Millipore

CellASIC ONIX2 Manifold XT (Temperature Controlled)

Synonym(s):

ONIX2 Manifold XT, Temperature Controlled Manifold

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$10,290.00

$10,290.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 EA
$10,290.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NC.51

$10,290.00


Check Cart for Availability

Request more information

General description

CellASIC® ONIX2 Manifold XT is a convective heat exchange module that aids in achieving the ideal temperature. It works by recirculating air gently and uniformly that warms or cools the cells.

Application

Research Category
Cell Culture

Features and Benefits

  • Ideal for small sized culture volume
  • Maintains a precise cell culture environment

Other Notes

When cleaning the Manifold XT tubing, keep the manifold right side up and level, and do not expose the heat exchange module to liquid.

Legal Information

CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Convert your microscope to a live cell imaging system – the CellASIC® ONIX2 system allows precise manipulation of the cell culture environment for dynamic live cell imaging and analysis using mammalian, yeast, and bacterial cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service