Skip to Content
Merck
All Photos(2)

Key Documents

MABC1142

Sigma-Aldrich

Anti-TIMP-4 Antibody, clone 9:4-7

clone 4-7, from mouse

Synonym(s):

Tissue inhibitor of metalloproteinases 4, Metalloproteinase inhibitor 4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$214.00
100 μL
$654.00

$214.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$214.00
100 μL
$654.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$214.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

4-7, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... TIMP4(7079)

General description

Tissue inhibitor of metalloproteinases 4 (UniProt: Q99727; also known as Metalloproteinase inhibitor 4, TIMP-4) is encoded by the TIMP4 gene (Gene ID: 7079) in human. The tissue inhibitors of metalloproteinases (TIMPs) represent a family of ubiquitous proteins which are natural inhibitors of the matrix metalloproteinases (MMPs). There are four known members of the TIMP family that are designated as TIMP-1, 2, 3, and 4. TIMP-4 is synthesized with a signal peptide (aa 1-29), which is cleaved off in the mature form. It has two MMP-binding regions (aa 30-33 and 99-100). TIMP-4 expressed is observed in heart, brain, pancreas, kidney, colon, testes, and adipose tissue. However, elevated levels of TIMP-4 have been reported in various cancers tissues, including breast, ovary, prostate, colon and kidney and are associated with poor survival prognosis. Hence, it can serve as a useful marker for disease progression. TIMP-4 is shown to complex MMPs and irreversibly inactivates them by binding to their catalytic zinc cofactor. This clone (9:4 7) is specific for human TIMP-4 and does not exhibit any cross-reactivity to other TIMPs. (Ref.: Gomez, DE., et al. (1997). Eur. J. Cell. Biol. 74(2); 111-122; Donover, PS et AL (2010). J. Cell. Biochem. 110(5); 1255-1261).

Specificity

Clone 9:4-7 specifically detects human TIMP-4.

Immunogen

GST-tagged full-length recombinant human TIMP-4.

Application

Anti-TIMP-4, clone 9:4-7, Cat. No. MABC1142, is a mouse monoclonal antibody that detects Tissue Inhibitor of Matrix Metalloproteinase 4 and has been tested for use in ELISA, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected TIMP-4 in human kidney tissue sections.

Immunohistochemistry (Paraffin) Analysis: A representative lot detected TIMP-4 in Immunohistochemistry applications (Donover, P.S., et. al. (2010). J Cell Biochem. 110(5):1255-61).

ELISA Analysis: A representative lot detected TIMP-4 in ELISA applications (Donover, P.S., et. al. (2010). J Cell Biochem. 110(5):1255-61).

Western Blotting Analysis: A representative lot detected TIMP-4 in Western Blotting applications (Donover, P.S., et. al. (2010). J Cell Biochem. 110(5):1255-61).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Immunohistochemistry (Paraffin) in human breast cancer tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected TIMP-4 in human breast cancer tissue sections.


Target description

25.50 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lieke Jäkel et al.
Acta neuropathologica communications, 12(1), 103-103 (2024-06-25)
Cerebral amyloid angiopathy (CAA) is a highly prevalent and progressive pathology, involving amyloid-β (Aβ) deposition in the cerebral blood vessel walls. CAA is associated with an increased risk for intracerebral hemorrhages (ICH). Insight into the molecular mechanisms associated with CAA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service