Skip to Content
Merck
All Photos(1)

Key Documents

ABE2867M

Sigma-Aldrich

Anti-NR5A2 (LRH1)

from rabbit

Synonym(s):

Nuclear receptor subfamily 5 group A member 2, Alpha-1-fetoprotein transcription factor, B1-binding factor, hB1F, CYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, LRH-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$252.00
100 μL
$586.00

$252.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
$252.00
100 μL
$586.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$252.00


Estimated to ship on27 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

technique(s)

ChIP: suitable (ChIP-seq)
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NR5A2(2494)

General description

Nuclear receptor subfamily 5 group A member 2 (UniProt: O00482; also known as Alpha-1-fetoprotein transcription factor, B1-binding factor, hB1F, CYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, LRH-1) is encoded by the NR5A2 (also known as B1F, CPF, FTF) gene (Gene ID: 2494) in human. It is a member of the nuclear receptor superfamily that binds to the sequence element 5′-AACGACCGACCTTGAG-3′ of the enhancer II of hepatitis B virus genes. It contains a DNA binding region (aa 83-104) and two NR C4-type zinc finger domains (aa 86-106 and 122-146). It is abundantly expressed in pancreas, but at reduced levels in liver, and at very low levels in heart and lung. It s expression has been demonstrated in the Hep-G2 cell line. Four different isoforms of LRH-1 have been described that are generated by alternate splicing. LRH-1 is reported to be involved in the regulation of expression of transcription factors, such as Oct4, HNF1, 3, and 4, which are known to participate in embryonic development. Its presence in high levels in beta-III Tubulin+ and NeuN+ neurons of the mantle zone (MZ) in early neural tube of mice (E10.5 and E12.5) indicate its involvement in neural differentiation. (Ref.: Stergiopoulos, A., and Politis, PK (2016). Nat. Commun. DOI: 10.1038/ncomms12230; Venteclef, N et al. (2008). J. Biol. Chem. 283(7); 3694 3701).

Specificity

This rabbit polyclonal antibody detects Nuclear receptor subfamily 5 group A member 2 (NR5A2) in human and murine cells.

Immunogen

Epitope: unknown
His-tagged full length recombinant human Nuclear receptor subfamily 5 group A member 2, isoform 2.

Application

Anti-NR5A2 (LRH1), Cat. No. ABE2867, is a highly specific rabbit polyclonal antibody that targets Nuclear receptor subfamily 5 group A member 2 and has been tested in Chromatin Immunoprecipitation (ChIP), Immunofluorescence, Immunoprecipitation, and Western Blotting.
Chromatin Immunoprecipitation Analysis: A representative lot detected NR5A2 (LRH1) in Chromatin Immunoprecipitation applications (Kaltezioti, V., et. al. (2010). PLoS Biol. 8(12):e1000565; Venteclef, N., et. al. (2010). Genes Dev. 24(4):381-95; Boulias, K., et. al. (2005). EMBO J. 24(14):2624-33; Chalkiadaki, A., et. al. (2005). Mol Cell Biol. 25(12):5095-105).

Western Blotting Analysis: A representative lot detected NR5A2 (LRH1) in Western Blotting applications (Chalkiadaki, A., et. al. (2005). Mol Cell Biol. 25(12):5095-105; Stergiopoulos, A., et. al. (2016). Nat Commun. 7:12230).

Immunofluorescence Analysis: A representative lot detected NR5A2 (LRH1) in Immunofluorescence YES titration paraffin sections from E10.5 (a), E12.5 (b), E14.5 (c) or E16.5 (d) mouse spinal cord (Stergiopoulos, A., et. al. (2016). Nat Commun. 7:12230).
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected NR5A2 (LRH1) in 10 µg of HEK293 cell lysate.

Target description

~68 kDa observed; 61.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service