Skip to Content
Merck
All Photos(1)

Key Documents

MAB8790

Sigma-Aldrich

Anti-HIV Antibody, p24, clone 6D4.1-10

clone 6D4.1-10, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$418.00

$418.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$418.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$418.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

6D4.1-10, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

Core protein p24.

Immunogen

Epitope: p24
HIV-1 viral lysate

Application

Anti-HIV Antibody, p24, clone 6D4.1-10 detects level of HIV & has been published & validated for use in ELISA & WB.
Immunoblotting, EIA

Optimal working dilutions must be determined by the end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Affinity purified. Liquid in PBS
Format: Purified

Storage and Stability

Maintain at 2-8°C

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

You Zhang et al.
The Journal of biological chemistry, 294(6), 2046-2059 (2018-12-20)
Insulin-induced gene 1 (INSIG1) regulates sterol synthesis by mediating the activation of sterol regulatory element-binding protein (SREBP) and the degradation of the HMG-CoA reductase (HMGCR). INSIG1 is up-regulated during HIV-1 infection, but its role in HIV-1 infection is unknown. In
Jing Ma et al.
The Journal of biological chemistry, 291(52), 26670-26685 (2016-10-28)
In the HIV-1 replication cycle, the endosomal sorting complex required for transport (ESCRT) machinery promotes viral budding and release in the late stages. In this process, the ESCRT proteins, ALIX and TSG101, are recruited through interactions with HIV-1 Gag p6.
Jing Lu et al.
Journal of medical virology, 93(6), 3607-3620 (2020-08-14)
Nutrient starvation is a common phenomenon that occurs during T cell activation. Upon pathogen infection, large amounts of immune cells migrate to infection sites, and antigen-specific T cells are activated; this is followed by rapid proliferation through clonal expansion. The

Questions

  1. Should the antibody be glycosylated for conjugation via the hydrazine method to nanoparticles?

    1 answer
    1. the glycosylation level of this antibody has not been characterized, but it should be intrinsically glycosylated.

      Hydrazine bioconjugation does not involve carbohydrates. It is accomplished via hydrazine aldehyde with the amino groups present on an antibody.

      However, carbohydrates on antibodies can be utilized via the periodate oxidation method for conjugation to nanoparticles.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service