Skip to Content
Merck
All Photos(1)

Key Documents

MAB858-4

Sigma-Aldrich

Anti-RSV Antibody, blend, clones 133-1H, 131-2G, and 130-12H

ascites fluid, Chemicon®, from mouse

Synonym(s):

RSV

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$688.00

$688.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$688.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$688.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

130-12H, monoclonal
131-2G, monoclonal
133-1H, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG

shipped in

wet ice

General description

RSV is a labile paramyxovirus that produces a characteristic fusion of human cells in tissue culture--the syncytial effect. Two subtypes, A and B, have been identified. Subtype B are characterized as the asymptomatic strains of the virus. The more severe clinical illnesses involve Subtype A strains.

Specificity

Blend of all RSV specifications of MAB858-1, 2 and 3. Recognizes the fusion, G, and nuclear protein of both A and B strains.

Immunogen

A2

Application

Anti-RSV Antibody, blend, clones 133-1H, 131-2G & 130-12H is an antibody against Respiratory Syncytial Virus for use in IF, ELISA.
ELISA at 1:800+

Indirect Immunofluorescence at 1:100-200+ (fresh frozen tissue sections)

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Liquid ascites fluid.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
RSV Control Slides, Catalogue Number 5012-5

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Talita Bianca Gagliardi et al.
Intervirology, 60(1-2), 56-60 (2017-09-05)
Syncytia formation is the hallmark of the cytopathic effect caused by human respiratory syncytial virus (HRSV), which is the most important viral respiratory pathogen in children. This article reports methodological improvements in primary HRSV isolation and the importance of syncytia
Tiina Talts et al.
Microbiology spectrum, 12(4), e0306723-e0306723 (2024-02-27)
Prevention of respiratory syncytial virus (RSV) infection is now a global health priority, with a long-acting monoclonal antibody and two RSV vaccines recently licenced for clinical use. Most licenced and candidate interventions target the RSV fusion (RSV-F) protein. New interventions
John P DeVincenzo et al.
The Journal of infectious diseases, 190(5), 975-978 (2004-08-06)
Premature infants and those with chronic lung disease or congenital heart disease are at high risk of severe respiratory syncytial virus (RSV) disease. Palivizumab (Synagis), a humanized anti-RSV monoclonal antibody, has been used extensively since 1998 to prevent severe RSV
Hayat Caidi et al.
Methods in molecular biology (Clifton, N.J.), 1442, 13-32 (2016-07-29)
Defective interfering viral particles have been reported as important determinants of the course of viral infection, and they can markedly temper the virulence of the infection. Here, we describe a simple method, based on limiting dilution, for the removal of
Ziyin Wang et al.
Mucosal immunology, 17(2), 272-287 (2024-02-22)
Respiratory viral infections remain a major cause of hospitalization and death worldwide. Patients with respiratory infections often lose weight. While acute weight loss is speculated to be a tolerance mechanism to limit pathogen growth, severe weight loss following infection can

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service