Skip to Content
Merck
All Photos(1)

Key Documents

MAB8262B-5

Sigma-Aldrich

Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H

clone 133-1H, Chemicon®, from mouse

Synonym(s):

RSV

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$2,150.00

$2,150.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1055

Request a Bulk Order

Select a Size

Change View
500 μG
$2,150.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$2,150.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZMS1055

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

133-1H, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

isotype

IgG2aκ

shipped in

wet ice

Specificity

Directed against the 47-49 kDa Fusion Protein of RSV. Antibody MAB858-1 recognizes both A and B RSV strains.

Immunogen

Respiratory Syncytial virus A2

Application

Detect Respiratory Syncytial Virus using this Anti-RSV Antibody, fusion protein, all type A, B strains, clone 133-1H validated for use in IF.
Immunofluorescence

Optimal working dilutions must be determined by the end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Biotin conjugated purified immunoglobulin. Liquid in 0.01M PBS, pH=7.1, 0.1% Sodium Azide with 15 mg/mL BSA as stabilizer

Storage and Stability

Store at 2 to 8ºC for up to 12 months from date of receipt. Protect from light.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megan Levy et al.
PLoS pathogens, 20(5), e1012198-e1012198 (2024-05-13)
Respiratory syncytial virus (RSV) is the most important viral agent of severe pediatric respiratory illness worldwide, but there is no approved pediatric vaccine. Here, we describe the development of the live-attenuated RSV vaccine candidate Min AL as well as engineered
N C Salisch et al.
NPJ vaccines, 4, 54-54 (2019-12-31)
Respiratory Syncytial Virus (RSV) can cause severe respiratory disease, yet a licensed vaccine is not available. We determined the immunogenicity of two homologous and one heterologous intramuscular prime-boost vaccination regimens using replication-incompetent adenoviral vectors of human serotype 26 and 35

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service