Skip to Content
Merck
All Photos(2)

Key Documents

MAB5790-I

Sigma-Aldrich

Anti-DSD-1 Antibody, clone 473HD

clone 473HD, from rat

Synonym(s):

DSD-1-proteoglycan

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$743.00

$743.00


Estimated to ship on09 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Estimated to ship on09 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

473HD, monoclonal

species reactivity

mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

General description

Anti-DSD-1 reacts with a glycosaminoglycan epitope selectively expressed on radial gliacytes of which a subpopulation shows stem cell properties. Cells isolated with this antibody from embryonic mouse tissue had multipotent characteristics and could turn into neurons, astrocytes and oligodendrocytes. The epitope is highly conserved and has been found in several vertebrates.

Immunogen

Purified tissue fraction corresponding to mouse DSD-1.

Application

Anti-DSD-1 Antibody, clone 473HD is an antibody against DSD-1, clone 473HD for use in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected DSD-1 in mouse hippocampus, mouse cerebellum, and mouse embryo tissue.

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected DSD-1 in 10 µg of mouse brain tissue lysate.

Target description

~400 kDa observed

Linkage

Replaces: MAB5790

Physical form

Format: Purified
Purified rat monoclonal IgMκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zaman Mirzadeh et al.
Nature metabolism, 1(2), 212-221 (2019-06-28)
In leptin-deficient ob/ob mice, obesity and diabetes are associated with abnormal development of neurocircuits in the hypothalamic arcuate nucleus (ARC)1, a critical brain area for energy and glucose homeostasis2,3. As this developmental defect can be remedied by systemic leptin administration

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service