Skip to Content
Merck
All Photos(1)

Key Documents

MAB570

Sigma-Aldrich

Anti-Heparin Antibody, clone A7.10

clone A7.10, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$725.00

$725.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$725.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$725.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A7.10, monoclonal

species reactivity

mouse

should not react with

pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
radioimmunoassay: suitable

isotype

IgG2b

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Hdgf(15191)

Specificity

Reacts with mouse heparin. Cross reacts with dextran sulfate by EIA. Also shows some cross reactivity to DNA and heparan sulfate.

Application

Detect Heparin using this Anti-Heparin Antibody, clone A7.10 validated for use in ELISA, RIA.
EIA/RIA (kd by liquid phase RIA for heparin is approximately

1.0 x 10-11 M.) Suggested concentration 1-10 μg/mL.

Inhibits the formation of heparin accelerated thrombin-antithrombin complex formation in vitro.

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Purified immunoglobulin. Liquid in 0.02M PB + 0.25M NaCl, pH 7.6 with 0.1% Sodium azide.

Storage and Stability

Maintain at 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service