Skip to Content
Merck
All Photos(1)

Key Documents

GF308

Sigma-Aldrich

NT-3 Protein, Human Recombinant Animal Free

Recombinant animal free human NT-3 is manufactured using all non-animal reagents.

Synonym(s):

NT3, Neurotrophin 3, HDNF, Nerve growth factor 2, Neurotrophic factor

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$707.00

$707.00


Estimated to ship on28 May 2025



Select a Size

Change View
10 μG
$707.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

$707.00


Estimated to ship on28 May 2025


recombinant

expressed in E. coli

Quality Level

Assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

UniProt accession no.

shipped in

dry ice

General description

Neurotrophin3 (NT3) is a member of the NGF family of neurotrophic factors (also named neurotrophins) that are required for the differentiation and survival of specific neuronal subpopulations in both the central as well as the peripheral nervous systems. The neurotrophin family is comprised of at least four proteins including NGF, BDNF, NT3, and NT4/5. NT-3 is expressed by neurons of the central nervous systems and can signal through the trk receptors. NT-3 promotes the growth and survival of nerve and glial cells. The amino acid sequences of human, murine and rat NT-3 are identical. Recombinant human NT-3 is a noncovalently linked homodimer, of two 13.6 kDa polypeptide monomers (240 total amino acid residues).Human and Mouse NT-3 sequences are identical.
Product Source: E. coli

Specificity

Cross Reactivty
None

Application

Recombinant animal free human NT-3 is manufactured using all non-animal reagents.
Research Category
Neuroscience
Research Sub Category
Growth Factors & Receptors

Quality

Bioactivity assay: The ED50 as determined by the dose-dependent induction of choline acetyl transferase activity in rat basal forebrain primary septal cell cultures was found in the range of 20-50 ng/ml.

Physical form

Product is filtered through a 0.2 micron filter before lyophilization. Product is presented in .
Recombinant animal free human NT-3 is manufactured using all non-animal reagents

Storage and Stability

Store at -20°C for up to 6 months from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Lawn et al.
The Journal of biological chemistry, 290(6), 3814-3824 (2014-12-30)
Neurotrophins and their receptors are frequently expressed in malignant gliomas, yet their functions are largely unknown. Previously, we have shown that p75 neurotrophin receptor is required for glioma invasion and proliferation. However, the role of Trk receptors has not been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service