Skip to Content
Merck
All Photos(1)

Key Documents

MAB3662

Sigma-Aldrich

Anti-TIF1 beta Antibody

ascites fluid, Chemicon®

Synonym(s):

TIF1beta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$660.00

$660.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$660.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$660.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

monoclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRIM28(10155)

Specificity

Recognizes transcriptional intermediary factor 1beta (TIF1beta). The epitope is localized between amino acids 123-834 of mouse TIF1beta. No cross reactivity with TIF1alpha.

Immunogen

Recombinant protein from mouse TIF1b.

Application

Anti-TIF1 beta Antibody is a Mouse Monoclonal Antibody for detection of Transcriptional Intermediary Factor 1 also known as TIF1 beta & has been validated in ELISA, WB, ICC.
Western blot: 1:500-1:5,000

Immunocytochemistry: 1:500-1:5,000

ELISA: 1:500-1:5,000

Optimal working dilutions must be determined by the end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Coluccio et al.
Epigenetics & chromatin, 11(1), 7-7 (2018-02-28)
The KZFP/KAP1 (KRAB zinc finger proteins/KRAB-associated protein 1) system plays a central role in repressing transposable elements (TEs) and maintaining parent-of-origin DNA methylation at imprinting control regions (ICRs) during the wave of genome-wide reprogramming that precedes implantation. In naïve murine
Mathieu Boulard et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(25), 14292-14298 (2020-06-12)
The mechanisms by which methylated mammalian promoters are transcriptionally silenced even in the presence of all of the factors required for their expression have long been a major unresolved issue in the field of epigenetics. Repression requires the assembly of
Nozomi Takahashi et al.
Genes & development, 33(1-2), 49-54 (2019-01-04)
Genomic imprinting is an epigenetic process regulated by germline-derived DNA methylation, causing parental origin-specific monoallelic gene expression. Zinc finger protein 57 (ZFP57) is critical for maintenance of this epigenetic memory during post-fertilization reprogramming, yet incomplete penetrance of ZFP57 mutations in
Helen M Rowe et al.
Development (Cambridge, England), 140(3), 519-529 (2013-01-08)
Endogenous retroviruses (ERVs) undergo de novo DNA methylation during the first few days of mammalian embryogenesis, although the factors that control the targeting of this process are largely unknown. We asked whether KAP1 (KRAB-associated protein 1) is involved in this
Maëva Devoucoux et al.
Molecular & cellular proteomics : MCP, 21(7), 100253-100253 (2022-06-01)
MRG15/MORF4L1 is a highly conserved protein in eukaryotes that contains a chromodomain (CHD) recognizing methylation of lysine 36 on histone H3 (H3K36me3) in chromatin. Intriguingly, it has been reported in the literature to interact with several different factors involved in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service