Skip to Content
Merck
All Photos(1)

Key Documents

MAB3516

Sigma-Aldrich

Anti-Heat Shock Protein 70 Antibody, clone W27

clone W27, Chemicon®, from mouse

Synonym(s):

HSP70

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
$743.00

$743.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
500 μL
$743.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$743.00


Estimated to ship on02 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

W27, monoclonal

species reactivity

bovine, human, monkey

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HSPA4(3308)

Specificity

Reacts with both the constitutive (HSP73) and inducible (HSP72) forms of HSP70.

POSITIVE CONTROL:

HeLa cells, BT474 cells or breast carcinoma.

Immunogen

HSP70 from HeLa cells.

Application

Immunoblotting: 0.5-1 μg/mL

Immunohistochemistry (frozen and formalin/paraffin): 2-4 μg/mL for 2 hours at room temperature. Staining of formalin fixed tissue sections requires boiling the tissue sections in 10mM citrate buffer, pH 6.0 for 10-20 minutes followed by cooling at room temperature for 20 minutes.

Immunoprecpitation: 2 μg/mg protein lysate (use Protein G)

Flow cytometry

Optimal working dilutions must be determined by end user.
This Anti-Heat Shock Protein 70 Antibody, clone W27 is validated for use in IP, WB, IH(P) for the detection of Heat Shock Protein 70.

Physical form

Format: Purified

Analysis Note

Control
POSITIVE CONTROL: BT474, MAD109, RenCa, SK-NEP-1, U937 cells or breast carcinoma.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chi Lam Au Yeung et al.
Nature communications, 7, 11150-11150 (2016-03-30)
Advanced ovarian cancer usually spreads to the visceral adipose tissue of the omentum. However, the omental stromal cell-derived molecular determinants that modulate ovarian cancer growth have not been characterized. Here, using next-generation sequencing technology, we identify significantly higher levels of
High dietary taurine reduces apoptosis and atherosclerosis in the left main coronary artery: association with reduced CCAAT/enhancer binding protein homologous protein and total plasma homocysteine but not lipidemia.
Zulli, A; Lau, E; Wijaya, BP; Jin, X; Sutarga, K; Schwartz, GD; Learmont, J; Wookey et al.
Hypertension null
Elisa Teyssou et al.
Neurobiology of aging, 58, 239-239 (2017-07-19)
Mutations in UBQLN2 have been associated with rare cases of X-linked juvenile and adult forms of amyotrophic lateral sclerosis (ALS) and ALS linked to frontotemporal dementia (FTD). Here, we report 1 known (c.1489C>T, p.Pro497Ser, P497S) and 3 novel (c.1481C>T, p.Pro494Leu
A Franchi et al.
Journal of cellular biochemistry, 121(4), 2877-2888 (2019-11-07)
Cells can communicate with other neighboring or distant cells through the secretion of extracellular vesicles (EV), composed of a lipid bilayer and bearing surface molecules that allow them to recognize target cells. In this way, EV induce signaling via different
Jeanette D Walton et al.
Neoplasia (New York, N.Y.), 6(6), 838-845 (2005-02-22)
Cellular heterogeneity is a hallmark of human neuroblastoma tumors and cell lines. Within a single neuroblastoma are cells from distinct neural crest lineages whose relative abundance is significant for prognosis. We postulate that a self-renewing multipotent tumor stem cell, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service