Skip to Content
Merck
All Photos(1)

Key Documents

MAB3108

Sigma-Aldrich

Anti-Maltose Binding Protein Antibody, clone 1H4

ascites fluid, clone 1H4, Chemicon®

Synonym(s):

MBP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$806.00

$806.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$806.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$806.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1H4, monoclonal

species reactivity

bacteria

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Escherichia coli K12 ... Male(948538)

Specificity

Maltose Binding Protein (MBP) and MBP fusion proteins.

Immunogen

MBP fusion protein

Application

Anti-Maltose Binding Protein Antibody, clone 1H4 detects level of Maltose Binding Protein & has been published & validated for use in ELISA, IP & WB.
ELISA: 1:1,000-1:5,000

Immunoblotting: 1:1,000-1:5,000

Immunoprecipitation: not tested.

Optimal working dilutions must be determined by the end user.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Physical form

Liquid, does not contain any preservative.

Storage and Stability

Maintain at -20°C in undiluted aliquots up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service