Skip to Content
Merck
All Photos(1)

Key Documents

SAB5500074

Sigma-Aldrich

Anti-CD8 antibody, Rabbit monoclonal

clone SP16, recombinant, expressed in proprietary host, tissue culture supernatant

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$469.00

$469.00


Check Cart for Availability


Select a Size

Change View
100 μL
$469.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$469.00


Check Cart for Availability

biological source

rabbit

Quality Level

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

SP16, monoclonal

species reactivity

human (tested)

technique(s)

immunohistochemistry: 1:100

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD8A(925)

General description

CD8 molecule consists of two chains, termed α and β chain, which are expressed as a disulphide-linked α/β heterdimer or as an α/α homodimer on T cell subset, thymocytes and NK cells. The majority of CD8+ T cells express CD8 as α/β heterdimer. CD8 functions as a coreceptor in concert with TCR for binding the MHC class I/peptide complex. The HIV-2 envelope glycoprotein binds CD8 α chain (but not β chain).

Immunogen

Synthetic peptide corresponding to C-terminus of alpha chain of the human CD8 molecule.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody supplied as tissue culture supernatant in TBS/1% BSA buffer pH 7.5 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuba N Gide et al.
Cancer cell, 35(2), 238-255 (2019-02-13)
Cancer immunotherapies provide survival benefits in responding patients, but many patients fail to respond. Identifying the biology of treatment response and resistance are a priority to optimize drug selection and improve patient outcomes. We performed transcriptomic and immune profiling on
Xue Wu et al.
Cancer science, 112(11), 4444-4456 (2021-08-13)
WEE1 plays an important role in the regulation of cell cycle G2/M checkpoints and DNA damage response (DDR). Inhibition of WEE1 can increase the instability of the genome and have anti-tumor effects in some solid tumors. However, it has certain
Barbara Ludwig et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(44), 11745-11750 (2017-10-29)
Transplantation of pancreatic islets for treating type 1 diabetes is restricted to patients with critical metabolic lability resulting from the need for immunosuppression and the shortage of donor organs. To overcome these barriers, we developed a strategy to macroencapsulate islets

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service