Skip to Content
Merck
All Photos(1)

Key Documents

MAB1393Z

Sigma-Aldrich

Anti-PECAM-1 Antibody, clone TLD-3A12, azide free

clone TLD-3A12, Chemicon®, from mouse

Synonym(s):

CD31, MAB1393

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$721.00

$721.00


Estimated to ship on02 June 2025



Select a Size

Change View
100 μG
$721.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$721.00


Estimated to ship on02 June 2025


biological source

mouse

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

TLD-3A12, monoclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

suitability

not suitable for immunohistochemistry (Paraffin)

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Related Categories

Specificity

Rat PECAM-1. Antibody gives positive reaction at high dilutions against recombinant rat PECAM-1, confirming specificity.

Immunogen

Activated, Lewis rat derived microglial cells.

Application

Anti-PECAM-1 Antibody, clone TLD-3A12, azide free detects level of PECAM-1 & has been published & validated for use in ELISA, FC, WB, IH.
Flow Cytometry: 1:50-1:100

Immunohistology: frozen sections (-20°C methanol fixation), not recommended for traditional formalin fixed tissues.

ELISA

Western Blotting: 1:500. MW = 130-140 kDa

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)

Physical form

Format: Purified
Purified immunoglobulin by affinity chromatography on Protein G. Liquid in phosphate buffered saline containing no preservatives.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: Activated microglial cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingchao Shi et al.
PLoS biology, 18(5), e3000705-e3000705 (2020-05-14)
Modeling the processes of neuronal progenitor proliferation and differentiation to produce mature cortical neuron subtypes is essential for the study of human brain development and the search for potential cell therapies. We demonstrated a novel paradigm for the generation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service