Skip to Content
Merck
All Photos(4)

Key Documents

SAB5700639

Sigma-Aldrich

Anti-CD31 antibody produced in rabbit

Synonym(s):

CD31, CD31/EndoCAM, GPIIA, PECA1, PECAM-1, PECAM1, endoCAM

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$790.00

$790.00


Check Cart for Availability


Select a Size

Change View
100 μL
$790.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$790.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

79
80
81
82

species reactivity

human, mouse, rat

concentration

3 mg/ml

technique(s)

immunofluorescence: 1:50-1:200
immunohistochemistry: 1:50-1:200
immunoprecipitation (IP): 1:50-1:200
western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... PECAM1(18613)

General description

The protein encoded by this gene is found on the surface of platelets, monocytes, neutrophils, and some types of T-cells, and makes up a large portion of endothelial cell intercellular junctions. The encoded protein is a member of the immunoglobulin superfamily and is likely involved in leukocyte migration, angiogenesis, and integrin activation.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 600 to the C-terminus of mouse CD31 (NP_001292086.1).

Physical form

PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Storage and Stability

Store at -20℃. Avoid freeze / thaw cycles.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service