HAWP04700F1
MCE Membrane Filter, 0.45 μm Pore Size
MF-Millipore™, filter diam. 47 mm, hydrophilic, white, pkg of 100 discs, F1
Synonym(s):
MF-Millipore™ Membrane Filter, 0.45 µm pore size
Select a Size
$492.00
Select a Size
About This Item
$492.00
Recommended Products
material
mixed cellulose esters (MCE) membrane
plain filter
white filter
Quality Level
sterility
non-sterile
feature
hydrophilic
packaging
pkg of 100 discs
manufacturer/tradename
MF-Millipore™
Millipore
sustainability
Greener Alternative Product
parameter
4 L/min-cm2 air flow rate
60 mL/min-cm2 water flow rate
75 °C max. temp.
diam.
47 mm
filter diam.
47 mm
thickness
150 μm
gravimetric extractables
2.5%
color
white
refractive index
n/D 1.51
matrix
MF-Millipore™
pore size
0.45 μm pore size
79 % porosity
bubble point
≥1.8 bar, air with water at 23 °C
greener alternative category
shipped in
ambient
General description
Application
Features and Benefits
- Applicable for environmental monitoring and biological applications.
- Available as packs of varying pore sizes, black or white color, with or without a gridded surface.
- Autoclavable and compatible with ethylene oxide and gamma irradiation sterilization.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service