Skip to Content
Merck
All Photos(1)

Key Documents

FCABS325PE

Sigma-Aldrich

Milli-Mark® Anti-acetyl-Histone H3-PE Antibody

Milli-Mark®, from rabbit

Synonym(s):

H3t, H3/t, H3/g, Histone H3.1t

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$589.00

$589.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$589.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$589.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

PE

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

Tetrahymena sp., human, mouse

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys9,Lys14)

Gene Information

human ... H3F3B(3021)

Related Categories

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Acetylation of histone H3 occurs at several different lysine positions in the histone tail and is performed by a family of enzymes known as Histone Acetyl Transferases (HATs). Acetylation of lysine14 is commonly seen in genes that are being actively transcribed into RNA.

Specificity

Antibody recognizes Tetrahymena acetyl Histone H3
This sequence is highly conserved and predicted to have broad species crossreactivity.

Immunogen

KLH-conjugated peptide corresponding to amino acids 1-20 of Tetrahymena histone H3 (ARTKQTAR[K*]STGG[K*]APRKQLC) where K* is acetylated

Application

This Milli-Mark Anti-acetyl-Histone H3-PE Antibody is validated for use in FC for the detection of acetyl-Histone H3.

Quality

Evaluated by flow cytometry using HeLa cells

Target description

15.5 kDa Calculated

Physical form

Purified rabbit polyclonal IgG conjugated to PE in PBS with 0.1% sodium azide and 15 mg/mL BSA

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HIV-1 expression within resting CD4+ T cells after multiple doses of vorinostat.
Archin, NM; Bateson, R; Tripathy, MK; Crooks, AM; Yang, KH; Dahl, NP; Kearney et al.
The Journal of Infectious Diseases null
Adam M Spivak et al.
Retrovirology, 13(1), 88-88 (2016-12-22)
Despite the durable viral suppression afforded by antiretroviral therapy, HIV-1 eradication will require strategies to target latently infected cells that persist in infected individuals. Protein kinase C (PKC) activation is a promising strategy to reactivate latent proviruses and allow for
Addie May I Nesbitt et al.
PloS one, 9(5), e94224-e94224 (2014-05-03)
Genetic, pharmacological, and environmental interventions that alter total levels of histone acetylation in specific brain regions can modulate behaviors and treatment responses. Efforts have been made to identify specific genes that are affected by alterations in total histone acetylation and
Valeria Barili et al.
Nature communications, 11(1), 604-604 (2020-02-01)
Hepatitis C virus infection (HCV) represents a unique model to characterize, from early to late stages of infection, the T cell differentiation process leading to exhaustion of human CD8+ T cells. Here we show that in early HCV infection, exhaustion-committed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service