Skip to Content
Merck
All Photos(1)

Key Documents

EZCURVCHA

Millipore

Power Supply for EZ-Pak® Dispenser Curve

External power supply/ charger for battery powered EZ-Pak® Dispenser Curve

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$415.00

$415.00


Available to ship on14 April 2025Details


Request more information

Select a Size

Change View
1 EA
$415.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.79

$415.00


Available to ship on14 April 2025Details


Request more information

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Pak®

output

65 W

input

sample type cosmetics
sample type bottled water
sample type pharmaceutical(s)
sample type beer

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

compatibility

for use with EZ-Pak® Dispenser Curve

shipped in

ambient

General description

The external power supply is an additional accessory of the EZ-Pak® Dispenser Curve. The EZ-Pak® Dispenser Curve revolutionizes filter handling for membrane filtration method testing by combining the benefits of high-speed sterile membrane dispensing, no-touch operation, and one-handed membrane transfer. The charger helps to power up the battery, and the dispenser can work with or without connection to a power outlet. A fully charged Lithium-ion battery can aid in dispensing up to 10,000 membranes. After 10 minutes of idle time, a standby mode is activated to save energy.

Application

The external power supply is an additional accessory of the EZ-Pak® Dispenser Curve used in General microbiological analysis of Beer, Bottled Water, Pharmaceutical Products Analysis, Environmental Analysis, Bioburden Testing, and Water Monitoring.

Legal Information

EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service