Skip to Content
Merck
All Photos(1)

Key Documents

DR1001

Sigma-Aldrich

Anti-8-Hydroxyguanosine Goat pAb

liquid, Calbiochem®

Synonym(s):

Anti-8-OHG, Anti-8-oxo-G

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$684.00

$684.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$684.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$684.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

goat

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity (predicted by homology)

all

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Goat polyclonal antibody supplied as undiluted serum. Recognizes 8-hydroxydeoxy-guanosine.
Recognizes 8-Hydroxydeoxyguanosine in Alzheimer′s disease brain tissue.
This Anti-8-Hydroxyguanosine Goat polyclonal antibody detects 8-Hydroxydeoxyguanosine in Alzheimer′s disease brain tissue. It is validated for use in ELISA and with paraffin sections.

Immunogen

8-Hydroxyguanosine

Application


ELISA (≥1:250,000)
Paraffin Sections (≥1:200, proteinase K pre-treatment recommended; see figure legend, see application references)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Alzheimer’s disease brain tissue

Other Notes

Abe, T., et al. 2003. Neurosci. Lett.336, 105.
Parker, A.R., et al. 2002. Cancer Res.62, 7230.
Does not cross-react with other naturally occurring nucleotides. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Gueven et al.
Cell death and differentiation, 14(6), 1149-1161 (2007-03-10)
Several different autosomal recessive genetic disorders characterized by ataxia with oculomotor apraxia (AOA) have been identified with the unifying feature of defective DNA damage recognition and/or repair. We describe here the characterization of a novel form of AOA showing increased
Joseph J Boyle et al.
The American journal of pathology, 174(3), 1097-1108 (2009-02-24)
Intraplaque hemorrhage accelerates atherosclerosis via oxidant stress and contributes to lesion development and destabilization. Normally, macrophages scavenge hemoglobin-haptoglobin (HbHp) complexes via CD163, and this process provokes the secretion of the anti-inflammatory atheroprotective cytokine interleukin (IL)-10. We therefore tested the hypothesis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service