Skip to Content
Merck
All Photos(4)

Key Documents

CYL4039

Sigma-Aldrich

PrecisION® hERG ion channel membrane preparation

Human ERG ion channel membrane preparation for Radioligand binding Assays.

Synonym(s):

Ion Channel Membrane Kit, hERG Channel Membrane Prep

Sign Into View Organizational & Contract Pricing

Select a Size

200 UNITS
$1,630.00

$1,630.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 UNITS
$1,630.00

About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.41

$1,630.00


Check Cart for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

manufacturer/tradename

PrecisION

UniProt accession no.

shipped in

dry ice

General description

Human ERG
The human ether-a-go-go related gene (hERG) is a potassium ion channel which is essential for normal cardiac repolarization. In drug screening models, the hERG K+ channel has been indicated to inhibit a wide variety of compounds, and its blockage can lead to cardiac QT interval prolongation and life threatening arrhythmias (Murphy et al. 2006). Cardiac safety relating to IKr K+ channels has become a major concern of regulatory agencies, as hERG channel inhibition has been identified as the firmest link to QT prolongation (Chiu et al. 2004). Millipore′s hERG membrane preparations are crude membrane preparations made from HEK293 stable recombinant cell lines (Millipore cat. # CYL3039), which are ideal HTS tools for screening antagonists against the hERG channel. The membrane preparations exhibit a Kd of 5.2 nM for [3H]-Astemizole. With 10 μg/well hERG Membrane Prep and 3.0 nM [3H]-Astemizole, a greater than 4-fold signal-to-background ratio was obtained.

Cell Line Description

GPCR Cell Lines

Application

Radioligand binding assay

Biochem/physiol Actions

Protein Target: hERG
Target Sub-Family: Potassium

Quality

Table 1. Signal: background and specific binding values obtained in a competition binding assay for the hERG membrane prep.
10 µg/well
Signal:Background 4.6
Specific Binding (cpm) 435


SPECIFICATIONS: 1 unit = 10 μg membrane preparation with [3H]-astemizole
Bmax with [3H]-astemizole: 6.7 pmol/mg
Kd for [3H]-astemizole: 5.2 nM

Table 2. Rank order comparison of various small molecule hERG inhibitors using [3H]-astemizole binding assay and E-Phys assays:
Binding Ki (nM) with hERG membrane preps Electrophysiology IC50 (nM) with hERG cell lines
Millipore CYL4039M Literature value* Millipore CYL4039 by IonWorks PPC Millipore CYL4039 by PatchExpressLiterature value by Patch Clamp
Astemizole 3.4 ± 0.7 3.3 ± 0.7 4.6 ± 0.003 ND 0.5
Dofetilide 5.8 ± 2.428 ± 6NDND15.3 ± 2.5
E-4031 6.1 ± 0.8 58 ± 5115 ± 0.02 144 ± 44 14
Pimozide 18.8 ± 2.3 14 ± 4NDND18
Cisapride 104.9 ± 30.9 123 ± 25 75.6 ± 0.02 5 ± 1 44.5 ± 10.6
Haloperidol 241.4 ± 21.4 234 ± 20 NDND63
Risperidone >4000 4302 ± 319NDND167
Verapimil >4000 3902 ± 529 1332 ± 1203100 ± 400 830

*Data obtained from Chiu et al., 2004


Specifications

Inucbation Conditions
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FB 96-well harvest plate (Millipore cat. # MAHF B1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with 25 mM Tris, pH 7.4, 130 mM NaCl, 5 mM KCl, 0.8 mM CaCl2, 0.1% BSA. Binding reaction is transferred to the filter plate, and washed 6 times (250μL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 10 mM Hepes, pH 7.4, 130 mM NaCl, 5 mM KCl, 0.8 mM MgCl2, 1 mM NaEGTA, 10 mM glucose, 0.1% BSA, filtered and stored at 4°C

Radioligand: [3H]-Astemizole (Perkin Elmer # NET-1140)

Wash Buffer: 25 mM Tris, pH 7.4, 130 mM NaCl, 5 mM KCl, 0.8 mM MgCl2, 0.05 mM CaCl2, 0.1% BSA, filtered and stored at 4°C


One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 4-fold signal: background with 3H-labeled Astemizole.

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membrane protein was adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at –70°C up to expiration date indicated on the label. Do not freeze and thaw.

Legal Information

Precision B+ is a registered trademark of Precision Scientific

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service