Skip to Content
Merck
All Photos(1)

Key Documents

CBL150-I

Sigma-Aldrich

Anti-CD3e Antibody, clone UCHT1

clone UCHT1, from mouse

Synonym(s):

T-cell surface glycoprotein CD3 epsilon chain, CD3-epsilon, CD3e, T-cell surface antigen T3/Leu-4 epsilon chain

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$479.00

$479.00


Estimated to ship on30 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1011

Request a Bulk Order

Select a Size

Change View
100 μG
$479.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$479.00


Estimated to ship on30 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1011

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

UCHT1, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CD3E(916)

General description

T-cell surface glycoprotein CD3 epsilon chain (UniProt P07766; also known as CD3-epsilon, CD3e, T-cell surface antigen T3/Leu-4 epsilon chain) is encoded by the CD3E (also known as T3E) gene (Gene ID 916) in human. The T cell receptor or TCR is a plasma membrane protein complex responsible for recognizing antigens bound to major histocompatibility complex (MHC) on antigen presenting cells. In 95% of T cells, the TCR is composed of a disulfide-linked alpha (α) and beta (β) dimer, while among 5% of T cells the TCR is composed of a gamma (γ) and a delta (δ) dimer. T cells are activated when their surface TCRs engage with antigen/MHC complex on antigen presenting cells. Each TCR alpha and beta chain contains a variable (V) and a constant (C) extracellular immunoglobulin-like domain, followed by a transmembrane region and a short cytoplasmic tail. The alpha and beta chains have very short cytoplasmic tail and rely on the CD3 (cluster of differentiation 3) co-receptor complex for transducing signaling further intracellularly. The CD3 co-receptor complex is composed of a CD3γ chain, a CD3δ chain, and two CD3ε chains. The CD3 complex, TCR dimer, and a ζ-chain together comprise the functional TCR complex. The CD3 epsilon chain is initially produced with a signal peptide (a.a. 1-22), the removal of which yields the mature chain (a.a. 23-207) with an Ig-like (a.a. 32-112) domain-containing extracellular region (a.a. 23-126), a transmembrane segment (a.a. 127-152), and a cytoplasmic tail (a.a. 153-207), which contains the immunoreceptor tyrosine-based activation motif or ITAM (a.a. 178-205) that is essential for mediating ZAP-70 interaction and transducing TCR signaling.

Specificity

Clone UCHT1 reacts with the extracellular domain of human CD3 epsilon chain (Clevers, H., et al. (1988). Eur. J. Immunol. 18(5):705-710).

Immunogen

Epitope: Extracellular domain.
Human infant thymocytes and lymphocytes from a patient with Sézary disease.

Application

Anti-CD3e Antibody, clone UCHT1 is an antibody against CD3e for use in Flow Cytometry, Immunoprecipitation, ELISA, Immunocytochemistry, Immunohistochemistry, Affects Function.
Immunoprecipitation Analysis: Representative lots immunoprecipitated CD3 epsilon-associated TCR complex (Schamel, W.W., et al. (2005). J. Exp. Med. 202(4):493-503; Poggi, A., et al. (1996). Int. Immunol. 8(12):1947-1953).
ELISA Analysis: A representative lot was used as the capture antibody for the detection of epsilon-associated TCR complex proteins by sandwich ELISA (Poggi, A., et al. (1996). Int. Immunol. 8(12):1947-1953).
Immunocytochemistry Analysis: A representative lot detected the exogensously expressed human CD3 epsilon chain on the surface of murine T cells by flourescent immunocytochemistry (Clevers, H., et al. (1988). Eur. J. Immunol. 18(5):705-710).
Immunocytochemistry Analysis: A representative lot was employed to determined the plasma membrane and cytoplasmic CD3 immunoreactivity in T cell populations and T-ALL blast cells by flourescent immunocytochemistry (Campana, D., et al. (1987). J. Immunol. 138(2):648-655).
Immunohistochemistry Analysis: A representative lot detected CD3+ T cells (T3+) in the periglandular stromal aggregates in frozen dometrium sections from female donors undergone routine dilation and curettage procedures (Kamat, B.R., and Isaacson, P.G. (1987). Am. J. Pathol. 127(1):66-73).
Affects Function: A representative lot induced surface IL-2R expression and proliferation of human peripheral blood T lymphocytes (Van Wauwe, J.P., et al. (1984). J. Immunol. 133(1):129-132).
Flow Cytometry Analysis: A representative lot was employed for fluorescence activated cell sorting (FACS) of CD3+ human lymphocytes (Beverley, P.C., and Callard, R.E. (1981). Science. 11(4):329-334).
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Quality

Evaluated by Flow Cytometry in human PBMC.

Flow Cytometry Analysis: 0.1 µg of this antibody detected CD3e in 1X10E6 human PBMC.

Target description

23.15 kDa (Proform) and 20.74 (mature form) calculated. ~27 kDa reported in UCHT1 immunoprecipitate (Poggi, A., et al. (1996). Int. Immunol. 8(12):1947-1953).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service