Skip to Content
Merck
All Photos(2)

Key Documents

MABF301

Sigma-Aldrich

Anti-Interleukin-7/IL-7 Antibody, clone 7B1.1

clone 7B1.1, from mouse

Synonym(s):

Interleukin-7, IL-7

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$705.00

$705.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$705.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$705.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7B1.1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IL7(3574)

Related Categories

General description

The protein named IL-7, or interleukin 7 is a hematopoietic growth factor secreted by stromal cell in the bone marrow and thymus but also produced by many other cell types including keratinocytes, epithelial cells, hepatocytes and macrophage/dendritic cells but not by classical lymphocytes. IL7 is a cytokine that has multiple functions and is critical for proper T cell maturation and lymphoid cell survival, and its expression stimulates the proliferation of B cells, T cell and NK cells. IL-7 is detected by cells via its receptor and binding results in a cascade of signals important for T cell development and for T cell survival within the periphery, as well as memory T cells. IL-7 expression is upregulated in various lymphomas and leukemias and because of its effects on T cell development IL-7 is being examined as a therapeutic agent for transplantation studies and even latent viral infection studies.

Immunogen

GST-tagged recombinant protein corresponding to human Interleukin-7/IL-7.

Application

Flow Cytometry Analysis: 1 µg of this antibody detected recombinant Interleukin-7/IL-7 in Jurkat T and Raji B lymphocytes.
This Anti-Interleukin-7/IL-7 Antibody, clone 7B1.1 is validated for use in western blotting & flow cytometry for the detection of Interleukin-7/IL-7.

Quality

Evaluated by Western Blotting in Interleukin 7 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected 0.5 µg of recombinant Interleukin-7/IL-7.

Target description

~ 17 kDa observed

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service