Skip to Content
Merck
All Photos(1)

Key Documents

CAX2-MBC20

Millipore

CellASIC ONIX2 Manifold Basic (No Temperature Control)

The CellASIC ONIX2 Manifold Basic seals to the CellASIC ONIX plates and delivers a defined gas environment to the cell culture region.

Synonym(s):

CellASIC Manifold, Manifold for CellASIC, ONIX2 Manifold Basic

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$2,330.00

$2,330.00


In StockDetails


Request more information

Select a Size

Change View
1 EA
$2,330.00

About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NC.51

$2,330.00


In StockDetails


Request more information

manufacturer/tradename

CellASIC® ONIX

shipped in

ambient

Application

Research Category
Cell Culture
The CellASIC ONIX2 Manifold Basic seals to the CellASIC ONIX plates and delivers a defined gas environment to the cell culture region.

Features and Benefits

  • Manifold seals the microfluidic plate via a one-touch vacuum mechanism
  • High precision at very small volumes due to the pressure-driven flow of liquids to the cell chambers
  • Prevents contamination between the flow system and the biological solutions
  • A low-profile manifold is easy to adapt to any inverted microscope stage

Legal Information

CELLASIC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Convert your microscope to a live cell imaging system – the CellASIC® ONIX2 system allows precise manipulation of the cell culture environment for dynamic live cell imaging and analysis using mammalian, yeast, and bacterial cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service