Skip to Content
Merck
All Photos(1)

Key Documents

BI0341

Sigma-Aldrich

Capping Reagent B

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
Pricing and availability is not currently available.

product line

Novabiochem®

Quality Level

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

manufacturer/tradename

Novabiochem®

shipped in

ambient

storage temp.

room temp

General description

Biosynthesis Novabiochem®

Application

BI0341-0201 is packagedin 24/400 glass bottles
DNA Reagents Label Change Explanation

Analysis Note

Assay (1-methylimidazole): 9.5-10.5% v/v
Assay (tetrahydrofuran): 89.0-91.0% v/v
Color (APHA): 10 max
Form: Clear liquid
Water (H2O): 300ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service