Skip to Content
Merck
All Photos(1)

Documents

BI0347

Sigma-Aldrich

Capping Reagent B1 40% Acetic Anhydride in Acetonitrile

Biosynthesis Novabiochem®

Synonym(s):

dna

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Acetylations
reagent type: derivatization reagent

transition temp

flash point 9.4 °C

shipped in

ambient

storage temp.

2-30°C

Application

BI0347-0200 is packaged in 24/400 glass bottles
BI0347-0505 is packaged in GL45 glass bottles
BI0347-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®

Analysis Note

Assay (acetic anhydride): 39.0-41.0% v/v
Assay (acetonitrile): 59.0-61.0% v/v
Color (APHA): 10 max
Component: Acetic Anhydride Purity: 99.0% min
Component: Acetonitrile Purity (By GC): 99.9% min
Form: Clear liquid
Water (H2O): 100ppm max

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.9 °F - closed cup

Flash Point(C)

9.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service