Skip to Content
Merck
All Photos(1)

Key Documents

AX0116

Supelco

Acetone

OmniSolv®

Synonym(s):

Dimethyl Ketone, 2-Propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
MDL number:
UNSPSC Code:
12352115
EC Index Number:
200-662-2
technique(s):
HPLC: suitable
LC/MS: suitable
application(s):
PFAS testing
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

Agency

suitable for DIN 38407-42
suitable for EPA ACB B21-02

Quality Level

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

product line

OmniSolv®

Assay

≥99.5% (corrected for water content, GC)

form

liquid

expl. lim.

13.2 %

technique(s)

HPLC: suitable
LC/MS: suitable

impurities

≤0.3 μeq/g Titrable acid
≤0.5% Water (H2O)
≤2 ng/L Capillary ECD responsive substances (as PCNB)
≤3 μg/L Capillary FID responsive substances (as decane)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

UV cutoff

≤329 nm

UV absorption

λ: 329 nm Amax: ≤1.00
λ: 330 nm Amax: ≤0.80
λ: 335 nm Amax: ≤0.30
λ: 340 nm Amax: ≤0.06
λ: 350 nm Amax: ≤0.005
λ: 400 nm Amax: ≤0.005

application(s)

PFAS testing

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

AX0116P-1 is packaged in poly-coated glass bottles
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .
OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC, corrected for water content)99.5% minCapillary ECD responsive substances (as PCNB)2ng/L maxCapillary FID responsive substances (as decane)3µg/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)1.0ppb maxInfrared SpectrumTo Pass TestRefractive index (at 20°C)Titrable acid0.3µeq/g maxUV Abs. at 329 nm1.00AU maxUV Abs. at 330 nm0.80AU maxUV Abs. at 335 nm0.30AU maxUV Abs. at 340 nm0.06AU maxUV Abs. at 350 nm0.005AU maxUV Abs. at 400 nm0.005AU maxUV Cut-off329nm maxWater (H2O)0.5% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Articles

An accurate and precise HPLC-PDA measurement of 32 cannabinoids and 2 cannflavins in hemp samples using a single extraction in less than 32 minutes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service