Skip to Content
Merck
All Photos(1)

Key Documents

AP200

Sigma-Aldrich

Goat Anti-Mouse light chain Antibody

1.3 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$328.00

$328.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$328.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$328.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

1.3 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

unmodified

Related Categories

General description

Antibody molecules typically comprise two immunoglobulin light chains covalently bound to a pair of heavy chains. Immunoglobulin light chains occur in two types, designated by the Greek letters kappa and lambda. Kappa and gamma light chains are approximately 250 amino acids in length with an average mass of about 25 kDa. The ratio of kappa to lambda found in the immunoglobulin population varies by species.

Specificity

Minimal cross-reaction with bovine, goat, horse, human, rabbit, rat, and sheep.
The antibody reacts strongly with native primary antibodies primarily with kappa light chains. It is not suitable for detecting lambda light chains. The antibody does not react with the heavy chain of mouse IgG. The antibody has been tested by ELISA and adsorbed to ensure minimal cross-reaction with bovine, goat, horse, human, rabbit, rat, and sheep immunoglobulins.

Immunogen

Epitope: Kappa light chain
Prepared from purified mouse IgG light chain.

Application

Detect Mouse light chain using this Goat anti-Mouse light chain Antibody validated for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Target description

25 kDa

Physical form

ImmunoAffinity Purified
Purified by immunoaffinity chromatography. Liquid in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6.

Storage and Stability

Maintain under sterile conditions at -20°C for up to twelve months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service