Key Documents
AIRGAPXL2
Milli-Q
Air Gap Device
For use with Milli-Q® CLX, HX, and HR 7000 series water purification systems.
About This Item
UNSPSC Code:
41104200
NACRES:
JA.13
Pricing and availability is not currently available. Contact Technical Service
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
General description
This accessory can be connected directly to a 10 mm OD tubing to provide protection from bacterial contamination on the reject tubing.
Application
For the bacteriological protection of reject lines.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany