Skip to Content
Merck
All Photos(1)

Key Documents

AG345

Sigma-Aldrich

Na+/H+ Exchanger-1 Peptide

control for AB3081/AB3082, liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
Product AG345 is not currently sold in your country. Contact Technical Service

Product Name

Na+/H+ Exchanger-1, Control Peptide for AB3081/AB3082,

biological source

rat

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

inhibition assay: suitable (peptide)

NCBI accession no.

UniProt accession no.

General description

Na+/H+ Exchanger 1 (NHE1) control peptide.

Application

Antibody blocking (AB3081/AB3082)

Optimal working concentration must be determined by the end user.

Physical form

Liquid in PBS, pH 7.5.

Storage and Stability

Store at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service