Skip to Content
Merck
All Photos(1)

Key Documents

13858-U

Supelco

Dowex 1X8 Ion Exchange Resin

200-400 mesh, matrix styrene divinylbenzene (gel)

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 KG
$3,400.00

$3,400.00


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
2.5 KG
$3,400.00

About This Item

UNSPSC Code:
41115710
NACRES:
SB.52

$3,400.00


Estimated to ship on28 March 2025


Request a Bulk Order

Product Name

Dowex 1X8 formate form, 200-400 mesh, 2.5kg, matrix styrene divinylbenzene (gel)

description

cross-linkage 8%

Quality Level

form

beads

technique(s)

LPLC: suitable

matrix

styrene divinylbenzene (gel)

matrix active group

quaternary ammonium

particle size

200-400 mesh

capacity

1.0 meq/mL

separation technique

anion exchange

General description

Dowex 1X8, a strongly basic anion exchange resin, has been used as an absorbent in solid phase extraction.[1]

Application

  • Dowex 1X8 formate form was used to remove neutral monosaccharides and hexosamines from hydrolysates using ion-exchange chromatography.[2]
  • Dowex 1X8 formate form was used to purify organic acids from lyophilized fractions.[3]

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Makatsori et al.
Journal of chromatography. B, Biomedical sciences and applications, 712(1-2), 23-29 (1998-08-11)
Serum and tissue specimens from healthy C57BI mice and from mice with Lewis' lung cancer after metastasis were analyzed for N-acetyl- and N-glycolylneuraminic acid by high-performance liquid chromatography. Both neuraminic acids were present, while N-glycolylneuraminic acid was the predominant sialic
Mohamed Koubaa et al.
Analytical biochemistry, 436(2), 151-159 (2013-02-13)
The tricarboxylic acid (TCA) cycle is involved in the complete oxidation of organic acids to carbon dioxide in aerobic cells. It not only uses the acetyl-CoA derived from glycolysis but also uses breakdown products of proteins, fatty acids, and nucleic
Ayşem Uzer et al.
Forensic science international, 174(2-3), 239-243 (2007-06-09)
The Meisenheimer anions formed from TNT in KOH solutions in alcohol or acetone were used in screening tests for TNT among possible nitro-explosives. The same reaction was used for the spectrophotometric assay of TNT in soil by CRREL (Cold Regions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service