Skip to Content
Merck
All Photos(2)

Key Documents

ABS2099

Sigma-Aldrich

Anti-STRN3

from rabbit

Synonym(s):

Cell cycle autoantigen SG2NA, S/G2 antigen

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... STRN3(29966)

General description

Striatin-3 (UniProt: Q13033; also known as Cell cycle autoantigen SG2NA, S/G2 antigen) is encoded by the STRN3 (also known as GS2NA, SG2NA) gene (Gene ID: 29966) in human. Striatin-3 is a calmodulin-binding protein that bonds calmodulin a calcium dependent manner and may function as scaffolding or signaling protein. Two isoforms of Striatin-3 have been reported that are produced by alternative splicing. Its calmoduln-binding region is located within amino acids 166-183. It also contains a caveolin-binding region (aa 71-79), six WD repeats, and a coiled coil region (aa 77-136). The coiled coil domain and the PP2A subunit can form a stable core complex with a 2:2 stoichiometry. The coiled coil domain of striatin-3 is a novel type of PP2A regulatory subunit that functions as a scaffold for the assembly of the STRIPAK complex. PP2A and certain kinases, such as germinal central kinase III (GCKIII) interact with striatin to form striatin-interacting phosphatase and kinase (STRIPAK) complex that are known to regulate multiple cellular events. Wild-type striatin-3 is shown to strongly suppress apoptosis in Jurkat cells induced by the GCKIII kinase MST3. However, mutant forms of striatin-3 fail to exhibit this effect. (Ref.: Lu, Q., et al. (2004). Proc. Natl. Acad. Sci. USA 101(49); 17126-17131; Chen, C., et al. (2014). J. Biol. Chem. 289(14); 9651-61).

Specificity

This rabbit polyclonal antibody detect Striatin-3 in human and murine cells. It targets an epitope within 17 amino acids from the N-terminal half.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal half of human Striatin-3. The immunogen sequence is conserved in both alpha and beta isoforms.

Application

Anti-STRN3, Cat. No. ABS2099, is a rabbit polyclonal antibody that detects Striatin-3 and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected STRN3 in HeLa cells.
Research Category
Signaling

Quality

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected STRN3 in 10 µg of NIH3T3 cell lysate.

Target description

~87 kDa observed; 87.21 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service