Skip to Content
Merck
All Photos(1)

Key Documents

ABS1513-I-AF488

Sigma-Aldrich

Anti-phospho-Ubiquitin Antibody (Ser65), Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

Ubiqutin, Ser65 phosphorylated, Ub, Ser65 phosphorylated

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$773.00

$773.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$773.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$773.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

guinea pig (based on 100% sequence homology), bovine (based on 100% sequence homology), hamster (based on 100% sequence homology), mouse (based on 100% sequence homology), sheep (based on 100% sequence homology), rat (based on 100% sequence homology), nonhuman primates (based on 100% sequence homology), horse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer65)

Gene Information

human ... UBB(7314)

General description

Ubiqutin (Ub) is initially produced as a 229-a.a. Polyubiquitin-B (UniProt P0CG47) precursor protein encoded by the UBB gene (Gene ID 7314) or a 685-a.a. Polyubiquitin-C precursor protein (UniProt P0CG48) encoded by the UBC gene (Gene ID 7316) in human. The C-termini of Polyubiquitin-B (Cys229) and Polyubiquitin-C (Val685) are post-translationally removed, followed by additional posttranslational cleavages to yield multiple copies of idential 76-amino acid ubiquitin (also known as high mobility group nonhistone protein HMG-20) molecules (three copies per Polyubiquitin-B and nine copies per Polyubiquitin-C). Covalent modifications of target proteins via different Ub Lys residues are well documented events during cellular DNA repair, ER-associated degradation (ERAD), transcription activation, lysosomal and proteasomal degradation. PINK1 (PTEN induced putative kinase 1) is a Ser/Thr kinase that specifically accumulates in depolarized mitochondria, where it acts as a positive regulator of parkin (Park2) E3 ubiquitin (Ub) ligase activity by phosphorylating Ub at Ser65 as well as the corresponding Ser residue in parkin N-terminal Ub-like (UBL) domain. Phosphorylated Ub interacts with phosphorylated parkin at its GINGO0 domain in an allosteric manner, inducing a conformation change and exposing parkin RING2 domain catalytic cysteine for full ligase activity. NMR based conformation study reveals that Ser65-phosphorylated Ub exists in two conformations in solution. The major conformation displays altered surface properties, while the second phosphoUb conformation exhibits a retracted C-terminal tail by two residues into the Ub core. Ub Ser65 phosphorylation has little effect toward E1-mediated E2 charging and mainly affects the discharging of E2 enzymes to form polyUb chains. In addition, the majority of deubiquitinases (DUBs), including USP8, USP15 and USP30, are shown to exhibit impaired activity against Lys63-linked poly-phosphoUb.

Specificity

Target specificity was verified using the unconjugated antibody (ABS1513-I). Only Ser65-phosphorylated ubiquitin (Ub) and ubiquitinated proteins (including oligoUb and polyUb) containing Ser65-phosphorylated ubiquitin were detected. This antibody exhibits no reactivity toward Ub or ubiquitinated proteins in the absence of Ub Ser65 phosphorylation.

Immunogen

KLH-conjugated linear peptide corresponding to the ubiquitin target region sequence containing phosphorylated Ser65.

Application

Detect ubiquitin Ser65 phosphorylation using this rabbit polyclonal Anti-phospho-Ubiquitin (Ser65) Antibody, Alexa Fluor 488 Conjugate, Cat. No. ABS1513-I-AF488, validated for use in Immunocytochemistry.
Research Category
Signaling
The unconjugated antibody (Cat. No. AB1513-I) is also available for Immunocytochemistry and Western Blotting applications.

Quality

Evaluated by Immunocytochemistry in PINK1-expressing HeLa cells with or without CCCP treatment.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected CCCP treatment-induced ubiquitin Ser65 phosphorylation in PINK1-expressing HeLa cells.

Target description

~17/8 kDa (dimer/monomer) and ~45-250 kDa observed. 8.565/17.112/25.66/34.21 kDa (mono-/di/tri-/tetra-ubiquitin) calculated. Bands ~45-250 kDa represent ubiquitinated proteins containing ser65-phosphorylated Ub. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in PBS with 1.5% BSA and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Fan Huang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 2055-2074 (2020-01-08)
In spinal cord ischemia-reperfusion (I/R) injury, large amounts of reactive oxygen species can cause mitochondrial damage. Therefore, mitophagy acts as the main mechanism for removing damaged mitochondria and protects nerve cells. This study aimed to illustrate the important role of
Matthew G Williamson et al.
Human molecular genetics, 32(18), 2808-2821 (2023-06-29)
Mutations in the Leucine-Rich Repeat Kinase 2 (LRRK2) gene have been identified as one of the most common genetic causes of Parkinson's disease (PD). The LRRK2 PD-associated mutations LRRK2G2019S and LRRK2R1441C, located in the kinase domain and in the ROC-COR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service