Skip to Content
Merck
All Photos(2)

Key Documents

ABS1022

Sigma-Aldrich

Anti-Bile salt-activated lipase Antibody

serum, from rabbit

Synonym(s):

Bile salt-activated lipase, BAL, Bile salt-stimulated lipase, BSSL, Bucelipase, Carboxyl ester lipase, Cholesterol esterase, Pancreatic lysophospholipase, Sterol esterase

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$504.00

$504.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$504.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$504.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CEL(1056)

General description

Bile salt-activated lipase (BAL) is a secreted digestive enzyme produced by the pancreas that works in concert with pancreatic lipase and Colipase to allow the digestion and use of dietary triglycerides. BAL is also expressed in breast tissues and, interestingly, in eosinophils. BAL also interacts with galectin-10 (Charcot-Leyden Crystal), a member of the Galectin family involved in T cell proliferation and lysophospholipase inhibition activity and is thought to play a role in eosinophil development as well.

Immunogen

Corresponding to human Bile salt-activated lipase.

Application

Anti-Bile salt-activated lipase Antibody | ABS1022 is an antibody against Bile salt-activated lipase Antibody for use in Western Blotting, Immunocytochemistry.
Immunocytochemistry Analysis: A representative lot detected Bile salt-activated lipase in HEK293T cells (Mark Lowe, University of Pittsburgh).

Quality

Evaluated by Western Blotting in mouse pancreas tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Bile salt-activated lipase in 10 µg of mouse pancreas tissue lysate.

Target description

~70, 90 and 110 kDa observed. Multiple bands can be observed due to different levels of glycosylation (Swan, J.S., et al. (1992). Biochem J. 283:119-122).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service