Skip to Content
Merck
All Photos(3)

Key Documents

840074C

Avanti

Soy PA

Avanti Research - A Croda Brand 840074C

Synonym(s):

L-α-phosphatidic acid (Soy) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$978.00

$978.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$978.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$978.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840074C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840074C

concentration

10 mg/mL (840074C-25mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is a diacyl-glycerophospholipid, with two fatty acids and a phosphate group that are covalently attached to a glycerol molecule through ester linkages.[1] PA is rich in soy lecithin.[2] PA is produced endogenously by the hydrolytic action of the enzyme phospholipase D.[2][3] It is also produced through phosphorylation of diacyl glycerol (DAG) by diacylglycerol kinases (DGKs).[3]

Application

Soy PA is suitable for use in validation of PA probe.[3]

Biochem/physiol Actions

Phosphatidic acid (PA) is present in biomembranes. It is the precursor for other phospholipids and acts as a signaling molecule.[2] PA is implicated in NAPDH oxidase (NOX) activity.[3]

Packaging

5 mL Clear Glass Sealed Ampule (840074C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The mitotic and metabolic effects of phosphatidic acid in the primary muscle cells of turbot (Scophthalmus maximus)
Wang T, et al.
Frontiers in Endocrinology, 9 (2018)
Diacylglycerol kinases terminate diacylglycerol signaling during the respiratory burst leading to heterogeneous phagosomal NADPH oxidase activation
Schlam D, et al.
Journal of biological and chemical chronicles, 288(32), 23090-23104 (2013)
Daniel Schlam et al.
The Journal of biological chemistry, 288(32), 23090-23104 (2013-07-03)
It is commonly assumed that all phagosomes have identical molecular composition. This assumption has remained largely unchallenged due to a paucity of methods to distinguish individual phagosomes. We devised an assay that extends the utility of nitro blue tetrazolium for
Phosphatidic acid enhances mTOR signaling and resistance exercise induced hypertrophy
Joy JM, et al.
Nutrition, 11(1), 29-29 (2014)
Michal Bohdanowicz et al.
Molecular biology of the cell, 24(11), 1700-1712 (2013-04-12)
Macrophages and dendritic cells continuously survey their environment in search of foreign particles and soluble antigens. Such surveillance involves the ongoing extension of actin-rich protrusions and the consequent formation of phagosomes and macropinosomes. The signals inducing this constitutive cytoskeletal remodeling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service