Skip to Content
Merck
All Photos(1)

Key Documents

ABN78B

Sigma-Aldrich

Anti-NeuN (rabbit) Antibody, Biotin Conjugate

from rabbit, biotin conjugate

Synonym(s):

NEUronal Nuclei, Neuronal Nuclei

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$530.00

$530.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$530.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$530.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

immunohistochemistry: suitable

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Rbfox3(52897)

General description

ABN78 is a rabbit polyclonal version of the Anti-NeuN, clone A60 (MAB377), NeuN is a highly characterized and cited mouse monoclonal antibody that specifically recognizes the DNA-binding, neuron-specific protein NeuN, which is present in most CNS and PNS neuronal cell types of all vertebrates tested. NeuN protein distributions are apparently restricted to neuronal nuclei, perikarya and some proximal neuronal processes in both fetal and adult brain although, some neurons fail to be recognized by NeuN at all ages: INL retinal cells, Cajal-Retzius cells, Purkinje cells, inferior olivary and dentate nucleus neurons, and sympathetic ganglion cells are examples. Immunohistochemically detectable NeuN protein first appears at developmental timepoints that correspond with the withdrawal of the neuron from the cell cycle and/or with the initiation of terminal differentiation of the neuron. Immunoreactivity appears around E9.5 in the mouse neural tube and is extensive throughout the developing nervous system by E12.5. Strong nuclear staining suggests a nuclear regulatory protein function; however, no evidence currently exists as to whether the NeuN protein antigen has a function in the distal cytoplasm or whether it is merely synthesized there before being transported back into the nucleus. No difference between protein isolated from purified nuclei and whole brain extract on immunoblots has been found.

Specificity

This antibody recognizes NeuN at the N-terminus. The unconjugated version if this antibody (ABN78) has been shown to work in human and rat

Immunogen

Epitope: N-terminus
GST-tagged recombinant protein corresponding to mouse NeuN.

Application

Anti-NeuN (rabbit) Antibody, Biotin Conjugate detects level of NeuN (rabbit) & has been published & validated for use in IH.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Immunohistochemistry in adult mouse brain tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected NeuN in adult mouse brain tissue.

Target description

The unconjugated parent antibody (Cat. No. ABN78) has an observed molecular weight at ~46/48 kDa An uncharacterized band appears at ~62 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal conjugated to Biotin in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Adult mouse brain tissue

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service