Skip to Content
Merck
All Photos(1)

Key Documents

ABN1488

Sigma-Aldrich

Anti-TIAM-1

from rabbit, purified by affinity chromatography

Synonym(s):

T-lymphoma invasion and metastasis-inducing protein 1, TIAM-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), rat (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TIAM1(7074)

General description

T-lymphoma invasion and metastasis-inducing protein 1 (UniProt Q13009; also known as TIAM-1) is encoded by the TIAM1 gene (Gene ID 7074) in human. TIAM-1 is a ubiquitously expressed guanine nucleotide exchange factor (GEF) that specifically activates the Rho-like GTPase Rac1 in response to growth factors and cell-substrate interaction. TIAM-1 associates with GTP-bound Ras and functions as an effector that mediates Rac activation by Ras. TIAM-1 promotes the formation of adherens junctions and the induction of an epithelial-like phenotype in mesenchymal cells, and is essential for the formation and maintenance of cadherin-based adhesions. Tiam1-knockdown in mice protects against Ras-induced skin carcinogenesis as a result of increased apoptosis and reduced proliferation/growth of keratinocytes. However, the fewer skin tumors developed progress more frequently to malignancy in Tiam1-knockout mice, indicating that loss of Tiam1 stimulates malignant conversion. Cellular TIAM-1 is targeted for proteasome-dependent degradation by the SCFbTrCP ubiquitin ligase in cooperation with casein kinase 1 and the MEK/ERK pathway upon mitogenic stimulation.

Specificity

This rabbit polyclonal antibody targets a C-terminal region of human and mouse TIAM-1 sequences reported by UniProt (Q13009 and Q60610).

Immunogen

KLH-conjugated linear peptide corresponding to a C-terminal region sequence of human TIAM-1.

Application

Anti-TIAM-1 Antibody, Cat. No. ABN1488, is a highly specific rabbit polyclonal antibody, that targets TIAM-1 and has been tested in Western Blotting.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in Neuro-2a cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected TIAM-1 in 10 µg of Neuro-2a mouse neuroblastoma cell lysate.

Target description

~190 kDa observed. 177.4/170.4 kDa (human isoform 1/2) and 177.4 kDa (mouse) calculated (Met1 removed). Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexia Hervieu et al.
Science signaling, 13(637) (2020-06-25)
Receptor tyrosine kinases (RTKs) are often overexpressed or mutated in cancers and drive tumor growth and metastasis. In the current model of RTK signaling, including that of MET, downstream phosphatidylinositol 3-kinase (PI3K) mediates both cell proliferation and cell migration, whereas

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service