Skip to Content
Merck
All Photos(1)

Key Documents

ST1070

Sigma-Aldrich

Anti-Tiam1 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-T-Cell Lymphoma Invasion and Metastasis 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$439.00

$439.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$439.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$439.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TIAM1(7074)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~200 kDa Tiam1 protein.
Recognizes the ~200 kDa Tiam1 protein in Jurkat and NIH3T3 cells.
This Anti-Tiam1 Rabbit pAb is validated for use in Immunoblotting for the detection of Tiam1.
Tiam1 (T-cell lymphoma invasion and metastasis 1) was originally identified by its ability to induce invasion by T-lymphoma cells when amplified. Tiam1 is a specific guanine nucleotide exchange factor (GEF) for the GTPase, Rac. By activating Rac1, Tiam1 regulates many cellular functions including actin cytoskeleton dynamics, cell proliferation, and cell survival. Tiam1 activity is regulated by its translocation to the membrane and the subsequent binding of its PH domain to phosphatidylinositol-3,4,5-triphosphate. Tiam1 is cleaved in a caspase-dependent manner in apoptotic cells, rendering it inactive as a result of an inability to translocate to the plasma membrane.

Immunogen

Human
a synthetic peptide corresponding to amino acids from the C-terminal domain of human Tiam1

Application

Immunoblotting (1:1000)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Highly Toxic (H)

Physical form

In Tris-Citrate/phosphate, pH 7.0-8.0.

Reconstitution

Following initial use, aliquot and freeze (-20°C) for long-term storage. Avoid freeze/thaw cycles.

Analysis Note

Positive Control
Jurkat or NIH3T3 cells

Other Notes

Antibody should be titrated for optimal results in individual systems.
Mertens, A.E., et al. 2003. FEBS Lett.546, 11.
Qi, H., et al. 2001. Cell Growth Differ.12, 603.
Stam, J.C., et al. 1997. J. Biol. Chem.272, 28447.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Axel Walch et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 21(5), 544-552 (2008-02-05)
Rho GTPases are a family of major regulators of E-cadherin-mediated cell adhesion that are implicated in the carcinogenic process by deregulated expression of the family members itself or of upstream modulators or downstream effectors. Combined investigation of the Rho GTPase
Elena A Afanasyeva et al.
Life science alliance, 4(5) (2021-03-05)
The migrational propensity of neuroblastoma is affected by cell identity, but the mechanisms behind the divergence remain unknown. Using RNAi and time-lapse imaging, we show that ADRN-type NB cells exhibit RAC1- and kalirin-dependent nucleokinetic (NUC) migration that relies on several
Ramesh Chandra et al.
Frontiers in molecular neuroscience, 6, 13-13 (2013-06-08)
Exposure to psychostimulants results in structural and synaptic plasticity in striatal medium spiny neurons (MSNs). These cellular adaptations arise from alterations in genes that are highly implicated in the rearrangement of the actin-cytoskeleton, such as T-lymphoma invasion and metastasis 1
Davide Gianni et al.
Molecular biology of the cell, 19(7), 2984-2994 (2008-05-09)
NADPH oxidase (Nox) family enzymes are one of the main sources of cellular reactive oxygen species (ROS), which have been shown to function as second messenger molecules. To date, seven members of this family have been reported, including Nox1-5 and
Ahmed T Kurdi et al.
Nature communications, 7, 13048-13048 (2016-10-12)
RORγt is a master transcription factor of Th17 cells and considered as a promising drug target for the treatment of autoimmune diseases. Here, we show the guanine nucleotide exchange factor, Tiam1, and its cognate Rho-family G protein, Rac1, regulate interleukin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service