Skip to Content
Merck
All Photos(1)

Key Documents

ABF125

Sigma-Aldrich

Anti-S100A8/MRP-8 Antibody (Azide Free)

serum, from rabbit

Synonym(s):

Protein S100-A8, Calgranulin-A, Chemotactic cytokine CP-10, Leukocyte L1 complex light chain, Migration inhibitory factor-related protein 8, MRP-8, p8, Pro-inflammatory S100 cytokine, S100 calcium-binding protein A8, S100A8/MRP-8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$537.00

$537.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$537.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$537.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

activity assay: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... S100A8(6279)

General description

S100A8, also known as Protein S100-A8, Calgranulin-A, Calprotectin L1L subunit, Cystic fibrosis antigen (CFAG), Leukocyte L1 complex light chain, Migration inhibitory factor related protein 8 (MRP-8), p8, S100 calcium binding protein A8, or Urinary stone protein band A, and encoded by the gene S100A8/CAGA/CFAG/MRP8, is an important protein that binds both zinc and calcium and plays an important role in the regulation of inflammatory processes and immune response. Found in complex with its partner S100A9, S100A8 facilitates chemotaxis, fatty acid trafficking, cytoskeleton reorganization, and NAPDPH oxidase activation and other intracellular activities, as well as a host of extracellular induced activities such as extracellular proinflammatory, antimicrobial, oxidant scavenging, and apoptotic activities . Additionally S100A8 can act as a potent autoimmunity amplifier and appears to augment various cancers and their spread when over expressed. S100A8 is widely expressed and used as biomarker in patients with inflammatory diseases and as a cancer marker in multiple forms of cancer.

Immunogen

Recombinant protein corresponding to mouse S100A8/MRP-8.

Application

Activity Assay Analysis: A representative lot detected that S100A8/MRP-8 reduces mouse neutrophil accumulation in dorsum-raised air pouchs in the presence of MSU crystals (Ryckman, C., et al. (2003). ARTHRITIS & RHEUMATISM. 48(8):2310–2320).
Immunohistochemistry Analysis: A representative lot detected S100A8/MRP-8 in mouse arthritic paw tissue (Cesaro, A., et al. (2012). PLOS ONE. 7(9):e4578).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-S100A8/MRP-8 Antibody (Azide Free) is validated for use in Western Blotting and Immunohistochemistry and Activity Assay for the detection of S100A8/MRP-8 (Azide Free).

Quality

Evaluated by Western Blotting in human spleen tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected S100A8/MRP-8 in 10 µg of human spleen tissue lysate.

Target description

~10 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal serum without preservatives.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service