Skip to Content
Merck
All Photos(7)

Documents

HPA007575

Sigma-Aldrich

Anti-S100A6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Calcyclin antibody produced in rabbit, Anti-Growth factor-inducible protein 2A9 antibody produced in rabbit, Anti-MLN 4 antibody produced in rabbit, Anti-PRA antibody produced in rabbit, Anti-Prolactin receptor-associated protein antibody produced in rabbit, Anti-Protein S100-A6 antibody produced in rabbit, Anti-S100 calcium-binding protein A6 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

AIFHKYSGREGDKHTLSKKELKELIQKELTIGSKLQDAEIARLMEDLDRNKDQEVNFQEYVTFLGALALIYNE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... S100A6(6277)

General description

S100A6 (S100 calcium binding protein A6) gene encodes an EF-hand calcium binding protein that is a member of the S100 family of proteins. It is also called calcyclin and contains a hydrophobic hinge region that connects the EF-hands. It is expressed in lineage restricted astrocyte precursors and may be useful in analyzing the characteristics of astrocyte precursors. It is also expressed in neural stem cells.

Immunogen

Protein S100-A6 recombinant protein epitope signature tag (PrEST)

Application

Anti-S100A6 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

S100A6 (S100 calcium binding protein A6) protein functions in processes such as cell proliferation and differentiation, calcium homeostasis, and neuronal degeneration in the brain. It may act as a buffering protein similar to calbindin. It may function in ubiquitinylated proteolytic degradation and cytoskeletal dynamics. Inhibition of this protein results in a decrease in the production of new astrocytes. S100A6 is involved in the differentiation and maturation of astrocytes.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70728

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barnita Haldar et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(2), 3179-3196 (2020-01-10)
ISOC is a cation current permeating the ISOC channel. In pulmonary endothelial cells, ISOC activation leads to formation of inter-endothelial cell gaps and barrier disruption. The immunophilin FK506-binding protein 51 (FKBP51), in conjunction with the serine/threonine protein phosphatase 5C (PPP5C)
Mehdi Damaghi et al.
Nature communications, 6, 8752-8752 (2015-12-15)
Early cancers are avascular and hence, profoundly acidic. Pre-malignant cells must adapt to acidosis to thrive in this hostile microenvironment. Here, we investigate MCF-7 cells that are adapted to grow in acidic conditions using SILAC proteomics and we reveal a
Jun Yamada et al.
Hippocampus, 24(1), 89-101 (2013-10-12)
S100A6 (calcyclin), an EF-hand calcium binding protein, is considered to play various roles in the brain, for example, cell proliferation and differentiation, calcium homeostasis, and neuronal degeneration. In addition to some limbic nuclei, S100A6 is distributed in the rostral migratory
Narendra Padhan et al.
Scientific reports, 7(1), 1490-1490 (2017-05-06)
Detection and quantification of proteins and their post-translational modifications are crucial to decipher functions of complex protein networks in cell biology and medicine. Capillary isoelectric focusing together with antibody-based detection can resolve and identify proteins and their isoforms with modest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service