Skip to Content
Merck
All Photos(1)

Key Documents

Z805939

Hellmanex III

Hellmanex™ III

Special Cleaning Concentrate for cuvettes

Synonym(s):

III-Grade Laboratory Detergent

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$167.00

$167.00


Estimated to ship on03 April 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
1 EA
$167.00

About This Item

UNSPSC Code:
12000000
NACRES:
SB.32

$167.00


Estimated to ship on03 April 2025


Request a Bulk OrderRequest more information

packaging

pkg of 1 L (PE bottle)

manufacturer/tradename

Hellma

technique(s)

UV/Vis spectroscopy: suitable
photometry: suitable

General description

Hellmanex III significantly reduces the surface tension of water. The removal of dirt particles is also assured by the good wetting action of a Hellmanex III aqueous solution, while its high emulsifying and dispersing capabilities prevent the redeposition of the loosened particles. Special surface-active substances facilitate the residue-free rinsing of the optical components once they have been cleaned.
Hellmanex III is filtered to 1 μm and is therefore virtually free of solid particles.

Application

The optimal dilution (usually 0.5 to 2 vol-%) depends on several factors, such as the hardness of the water, the degree and type of contamination, the temperature, etc. The use of demineralised water improves the cleaning characteristics.
An increase in temperature speeds up the cleaning process. However, at high temperatures it is necessary to avoid thermal shock. The cells should be pre-warmed before being submerged into hot cleaning solutions.
Hellmanex III, when diluted to its working concentration of 2%, is suitable for use with glass, quartz, sapphire, porcelain, ceramics, plastics, and ferrous metals. It can also be used, with some limitations, on metals that are sensitive to corrosion such as aluminium and other non-ferrous metals. In such cases, avoid prolonged application. In the case of materials not listed here, suitability tests should be carried out or enquiries directed to Hellma.
The concentrate can be stored in vessels made of stainless steel, PE, PP, PVC, FPM, or NBR.

Legal Information

Hellmanex is a trademark of Hellma GmbH

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Niekamp et al.
The EMBO journal, 38(13), e101414-e101414 (2019-07-04)
The movement of a molecular motor protein along a cytoskeletal track requires communication between enzymatic, polymer-binding, and mechanical elements. Such communication is particularly complex and not well understood in the dynein motor, an ATPase that is comprised of a ring
Doug K Tischer et al.
eLife, 8 (2019-04-06)
T cells are thought to discriminate self from foreign peptides by converting small differences in ligand binding half-life into large changes in cell signaling. Such a kinetic proofreading model has been difficult to test directly, as existing methods of altering
Stefan Niekamp et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(10), 4275-4284 (2019-02-17)
Light microscopy is a powerful tool for probing the conformations of molecular machines at the single-molecule level. Single-molecule Förster resonance energy transfer can measure intramolecular distance changes of single molecules in the range of 2 to 8 nm. However, current
Sudha Kumari et al.
The EMBO journal, 39(5), e102783-e102783 (2020-01-03)
When migratory T cells encounter antigen-presenting cells (APCs), they arrest and form radially symmetric, stable intercellular junctions termed immunological synapses which facilitate exchange of crucial biochemical information and are critical for T-cell immunity. While the cellular processes underlying synapse formation

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service