Skip to Content
Merck
All Photos(2)

Key Documents

ABE1844

Sigma-Aldrich

Anti-NRF-1 Antibody

from rabbit

Synonym(s):

Nuclear respiratory factor 1, NRF-1, Alpha palindromic-binding protein, Alpha-pal, NRF-1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μG
$702.00

$702.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
200 μG
$702.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$702.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology), horse (based on 100% sequence homology), bovine (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

ChIP: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Nrf1(18181)

General description

Nuclear respiratory factor (UniProt Q9WU00; also known as Alpha-pal, Alpha palindromic-binding protein, NRF-1) is encoded by the Nrf1 gene (also known as C87038, D6Ertd415e) in murine species (Gene ID 18181). NRF-1 is a redox-responsive transcription factor that plays an anti-oxidant, pro-survival role in mitochondrial biogenesis process. NRF-1 is upregulated and activated/phosphorylated by Akt in response to oxidant exposure or increased cellular reactive oxygen species (ROS). This results in NRF-1 nuclear translocation, where it binds to DNA as a homodimer and mediates the transcription of mitochondrial transcription factor A (mtTFA) that is needed for mitochondrial DNA transcription.

Immunogen

KLH-conjugated linear peptide corresponding to mouse NRF-1.

Application

Immunocytochemistry Analysis: A representative lot detected NRF-1 in neonatal rat cardiomyoblasts (H9c2) (Data courtesy of Hagir B. Suliman and Claude A. Piantadosi , Duke University Medical Center).
Western Blotting Analysis: A representative lot detected NRF-1 in rat hepatoma cell line H4IIE treated with t-BH (Suliman, H.B., et al. (2003). J. Biol. Chem. 278(42):41510-41518).
Western Blotting Analysis: A representative lot detected NRF-1 in mouse hepatic nuclear proteins (Cherry, A.D., et al. (2014). J. Biol. Chem. 289(1):41-52).
Western Blotting Analysis: A representative lot detected NRF-1 in mouse liver after S. aureus inoculation (MacGarvey, N.C., et al. (2012). Am J Respir Crit Care Med. 185(8):851-861).
Western Blotting Analysis: A representative lot detected NRF-1 in rat hepatocytes and rat hepatoma cell line H4IIE cells (Carraway, M.S., et al. (2010). Circ Res. 106(11):1722-1730).
Chromatin Immunoprecipitation Analysis: A representative lot detected enhanced NRF-1 occupancy at the Sod2 promoter region in liver nuclear extracts from mice during S. aureus sepsis (Cherry, A.D., et al. (2014). J. Biol. Chem. 289(1):41-52).
Immunocytochemistry Analysis: A representative lot detected NRF-1 in Staphylococcus aureus peptidoglycan
(PGN) or dichloromethane (DCM)/carbon monoxide (CO) treated HepG2 cells (MacGarvey, N.C., et al. (2012). Am J Respir Crit Care Med. 185(8):851-861).
Immunocytochemistry Analysis: A representative lot detected NRF-1 in H9C2 rat cardiomyocytes (Carraway, M.S., et al. (2010). Circ Res. 106(11):1722-1730).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
This Anti-NRF-1 Antibody is validated for use in Western Blotting, Immunocytochemistry, Chromatin Immunoprecipitation (ChIP) for the detection of NRF-1 .

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected NRF-1 in 10 µg of human liver tissue lysate.

Target description

~54/58 kDa observed

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal antibody in buffer containing PBS and 0.01% Sodium Azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service