Skip to Content
Merck
All Photos(2)

Key Documents

860620P

Avanti

Lyso SM (dihydro) (d18:0)

Avanti Research - A Croda Brand, powder

Synonym(s):

Sphinganine Phosphorylcholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$1,320.00

$1,320.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$1,320.00

About This Item

Empirical Formula (Hill Notation):
C23H51N2O5P
CAS Number:
Molecular Weight:
466.64
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,320.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

1 ea of 5 mg (860620P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

deuterated lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](CCCCCCCCCCCCCCC)(O)[C@@]([H])(N)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C23H51N2O5P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(26)22(24)21-30-31(27,28)29-20-19-25(2,3)4/h22-23,26H,5-21,24H2,1-4H3/t22-,23+/m0/s1

InChI key

GSEOJHIBPQRSNH-XZOQPEGZSA-N

General description

Lyso-dihydro-Sphingomyelin (lyso-dihydro-SM) is an analog of Lyso sphingomyelin (Lyso-SM).[1] Lyso-SM is a derivative of sphingomyelin.[2]

Biochem/physiol Actions

Lyso sphingomyelin (Lyso-SM) is involved in cellular functioning of heart, brain, blood vessels and skin. It also acts as a lipid mediator.[3]

Packaging

5 mL Clear Glass Sealed Ampule

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingosylphosphocholine reduces the calcium ion requirement for activating tissue transglutaminase
Lai T S, et al.
The Journal of biological chemistry, 272(26), 16295-16300 (1997)
Lyso-sphingomyelin is elevated in dried blood spots of Niemann-Pick B patients
Chuang W L, et al.
Molecular Genetics and Metabolism, 111(2), 209-211 (2014)
A novel enzyme that cleaves the N-acyl linkage of ceramides in various glycosphingolipids as well as sphingomyelin to produce their lyso forms.
Ito M
The Journal of Biological Chemistry, 270(41), 24370-24374 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service