Skip to Content
Merck
All Photos(2)

Key Documents

ABC131

Sigma-Aldrich

Anti-Ambra1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Activating molecule in BECN1-regulated autophagy protein 1, Ambra, Ambra-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$660.00

$660.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$660.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$660.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AMBRA1(55626)

General description

Activating molecule in BECN1-regulated autophagy protein 1 (AMBRA1) is a developmental protein that is involved in autophagy, differentiation and nuerogenesis. AMBRA1 regulates autophagy and development of the nervous system, normal cell survival and proliferation, and the control of protein turnover during neuronal development.

Specificity

This antibody recognizes the C-terminus of Ambra1. This antibody should recognize isoforms 1 through 5 based upon immunogen design.

Immunogen

Epitope: C-terminus
GST-tagged recombinant protein corresponding to the C-terminus of mouse Ambra1.

Application

Anti-Ambra1 Antibody is a rabbit polyclonal antibody, validated for use in western blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
Western Blotting Analysis: 0.5 µg/mL of a representative lot detected Ambra1 in 10 µg of HeLa cell lysate.
Western Blotting Analysis: 1 µg/mL of a representative lot detected Ambra1 in human cell lysates (HeLa & U87MG) and in mouse tissue homogenates (liver, kidney, and embryo), as well as exogenously expressed myc-Ambra1 in lysates from transfected HEK293 cells. (Courtesy of Dr. Sabrina Di Bartolomeo, Cecconi Lab at the Dulbecco Telethon Institute, Italy).
Western Blotting Analysis: Target specificity of a representative lot was confirmed by Western blotting analysis of Ambra1-knock mouse embryos and HeLa cells treated with Ambra1 siRNA. (Courtesy of Dr. Sabrina Di Bartolomeo, Cecconi Lab at the Dulbecco Telethon Institute, Italy).
Western Blotting Analysis: A representative lot detected decreased Ambra1 expression in tissue homogenates (kidney, liver, and lung) from Ambra1+/- mice when compared with tissue samples from wild-type mice. (Cianfanelli, V., et al. (2015). Nat. Cell Biol. 17(1):20-30).

Quality

Evaluated by Western Blotting in MCF7 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Ambra1 in 10 µg of MCF7 cell lysate.

Target description

~140 kDa observed. Uniprot describes 7 isoforms at 143 kDa (isoform 1), 133 kDa (isoform 2), 87 kDa (isoform 3), 84 kDa (isoform 4), 100 kDa (isoform 5), 136 kDa (isoform 6), and 130kDa (Isoform 7). Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MCF7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angelica Jardim Costa et al.
Pharmaceutics, 13(2) (2021-01-27)
The pharmacological modulation of autophagy is considered a promising neuroprotective strategy. While it has been postulated that lithium regulates this cellular process, the age-related effects have not been fully elucidated. Here, we evaluated lithium-mediated neuroprotective effects in young and aged
Matteo Tiberti et al.
Cell death & disease, 13(10), 872-872 (2022-10-16)
Cancer genomics and cancer mutation databases have made an available wealth of information about missense mutations found in cancer patient samples. Contextualizing by means of annotation and predicting the effect of amino acid change help identify which ones are more
Francesca Nazio et al.
Acta neuropathologica, 142(3), 537-564 (2021-07-25)
Medulloblastoma (MB) is a childhood malignant brain tumour comprising four main subgroups characterized by different genetic alterations and rate of mortality. Among MB subgroups, patients with enhanced levels of the c-MYC oncogene (MBGroup3) have the poorest prognosis. Here we identify
Alessandra Romagnoli et al.
Cell death & disease, 14(8), 505-505 (2023-08-06)
Mycobacterium tuberculosis (Mtb) is known to evade host immune responses and persist in macrophages for long periods. A mechanism that the host uses to combat Mtb is xenophagy, a selective form of autophagy that targets intracellular pathogens for degradation. Ubiquitination
Manuela Antonioli et al.
Autophagy, 17(10), 2842-2855 (2020-11-12)
Oropharyngeal squamous cell carcinoma (OPSCC) is an increasing world health problem with a more favorable prognosis for patients with human papillomavirus (HPV)-positive tumors compared to those with HPV-negative OPSCC. How HPV confers a less aggressive phenotype, however, remains undefined. We

Articles

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service