Skip to Content
Merck
All Photos(1)

Documents

AB5898

Sigma-Aldrich

Anti-Protein Gene Product 9.5 Antibody

serum, Chemicon®

Synonym(s):

PGP 9.5, UCH-L1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

guinea pig

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... UCHL1(7345)
mouse ... Uchl1(22223)
rat ... Uchl1(29545)

Specificity

Recognizes Protein Gene Product 9.5 (PGP9.5).

Immunogen

A 17 amino acid peptide corresponding to amino acids 175-191 from soluble cytoplasmic human PGP9.5.

Application

Anti-Protein Gene Product 9.5 Antibody detects level of Protein Gene Product 9.5 & has been published & validated for use in IH.
Immunohistochemistry: 1:500.

Optimal working dilutions must be determined by end user.

blocking buffer for 1 hour at room temperature. Primary antibody was diluted in blocking buffer to the appropriate working dilution. Blocking buffer was removed and the slides were then incubated at 4°C for 18-24 hours with AB5898 (1:500). After rinsing in PBS 3 times sections were incubated for 60 minutes at room temperature with Cy3-conjugated secondary antibodies. After mounting in a mixture of PBS and glycerol (1:3) containing 0.1% p-phenylenediamine, sections were examined with a Nikon Microphot-SA epifluorescence microscope.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Physical form

Liquid. Contains 0.05% sodium azide.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Bagyánszki et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 23(6), e237-e248 (2011-04-08)
There are indications that alterations in the nitric oxide (NO) system of relaxation mediate gastrointestinal motor disturbances induced by chronic alcohol consumption (CAC). As CAC is known to inhibit the motility of the mouse small intestine, we investigated in this
Rebecca S Hornung et al.
Molecular pain, 18, 17448069211069255-17448069211069255 (2022-01-19)
Orofacial pain disorders are predominately experienced by women. Progesterone, a major ovarian hormone, is neuroprotective and antinociceptive. We recently reported that progesterone attenuates estrogen-exacerbated orofacial pain behaviors, yet it remains unclear what anatomical substrate underlies progesterone's activity in the trigeminal
Allison Doyle Brackley et al.
The Journal of biological chemistry, 292(21), 8762-8772 (2017-04-07)
μ-Opioid receptor (MOR) agonists are often used to treat severe pain but can result in adverse side effects. To circumvent systemic side effects, targeting peripheral opioid receptors is an attractive alternative treatment for severe pain. Activation of the δ-opioid receptor
Birol Yüce et al.
Gastroenterology, 133(2), 574-586 (2007-08-08)
Nociceptin is the endogenous agonist of the "orphan" opioid receptor-1 (ORL-1). We investigated whether activation of the ORL-1 receptor influences smooth muscle contractility and enteric neurotransmission within ascending myenteric reflex pathways of rats. Reverse transcriptase polymerase chain reaction was performed
Yanyan Xing et al.
The Journal of investigative dermatology, 141(5), 1308-1316 (2020-10-23)
Diverse sensory neurons exhibit distinct neuronal morphologies with a variety of axon terminal arborizations subserving their functions. Because of its clinical significance, the molecular and cellular mechanisms of itch are being intensely studied. However, a complete analysis of itch-sensing terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service