Skip to Content
Merck
All Photos(1)

Key Documents

AB5852

Sigma-Aldrich

Anti-SHANK1a Antibody, NT

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$602.00

$602.00


Estimated to ship on14 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$602.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$602.00


Estimated to ship on14 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Shank is a novel family of postsynaptic density proteins (PSD) that bind to PDS-95-associated protein GKAP. Its putative function is to act as a scaffold protein in the PSD to cross-link NMDA receptors/PSD-95 complexes and couple them to regulators of the actin cytoskeleton. Shank1a is a splice variant of Shank1.

Specificity

Recognizes Shank1a, N-terminal.

Immunogen

Epitope: N-terminus
Synthetic peptide corresponding to the N-terminus of rat Shank1a.

Application

Anti-SHANK1a Antibody, N-terminus is an antibody against SHANK1a for use in WB.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology
Western blotting: 1:500-1:1,000

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ofer Zohar et al.
Journal of immunology (Baltimore, Md. : 1950), 180(10), 6447-6451 (2008-05-06)
MHC class I molecules (MHC-I) have been implicated in nervous system development in the mouse. In this study we present evidence for the interaction of MHC-I with the NK cell receptor Ly49 in primary cortical neuronal cultures. We show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service