Skip to Content
Merck
All Photos(1)

Documents

407950

Sigma-Aldrich

Ionomycin free acid

from Streptomyces conglobatus, ≥90% (HPLC), waxy solid, Ca2+ mobile ion carrier, Calbiochem®

Synonym(s):

Ionomycin, Free Acid, Streptomyces conglobatus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H72O9
CAS Number:
Molecular Weight:
709.00
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

product name

Ionomycin, Free Acid, Streptomyces conglobatus, Ionomycin, Free Acid, Streptomyces conglobatus, CAS 56092-81-0, is a highly specific for divalent cations and more effective than A23187 (Cat. No. 100105) as a mobile ion carrier for Ca2+.

Quality Level

Assay

≥90% (HPLC)

form

waxy solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 10 mg/mL
methanol: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C41H72O9/c1-25(21-29(5)34(43)24-35(44)30(6)22-27(3)20-26(2)14-15-38(46)47)12-11-13-28(4)39(48)31(7)36(45)23-33-16-18-41(10,49-33)37-17-19-40(9,50-37)32(8)42/h11,13,24-33,36-37,39,42-43,45,48H,12,14-23H2,1-10H3,(H,46,47)/b13-11+,34-24-/t25-,26-,27+,28-,29-,30+,31+,32-,33?,36+,37?,39-,40+,41+/m1/s1

InChI key

PGHMRUGBZOYCAA-OJFQOPKISA-N

General description

Absorption max. (280 nm, MeOH): 18,600 M-1cm-1.
Highly specific for divalent cations (Ca2+ > Mg2+ >> Sr2+ = Ba2+). More effective than antibiotic A23187 as a mobile ion carrier for Ca2+. Complexes with Ca2+ between pH 7 and 9.5, causing significant absorption in the UV range; however, in contrast to A23187 (Cat. No. 100105), it is not fluorescent. Useful for studies of Ca2+ transport across biological membranes and measurement of cytoplasmic free Ca2+. Induces apoptotic neuronal degeneration in embryonic cortical neurons. Also induces cell cycle arrest in the G1 interval of mature Burkitt lymphoma cell lines.
Highly specific for divalent cations and more effective than A23187 (Cat. No. 100105) as a mobile ion carrier for Ca2+. Complexes with Ca2+ between pH 7 and 9.5. In contrast to A23187, it is not fluorescent. Useful for studies of Ca2+ transport across biological membranes and measurement of cytoplasmic free Ca2+. Induces apoptotic neuronal degeneration in embryonic cortical neurons and cell cycle arrest in the G1 phase in mature Burkitt lymphoma cell lines.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Mobile ion carrier for Ca2+
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 1 year at 4°C.

Other Notes

Aagaard-Tillery, K.M., and Jelinek, D.F. 1995. J. Immunol.155, 3297.
Takei, N., and Endo, Y. 1994. Brain Res. 652, 65.
Dupuy d’Angeac, A., et al. 1993. J. Cell Physiol.154, 44.
Lee, H.C. 1993. J. Biol. Chem.268, 293.
Kuznetsov, G., et al. 1992. J. Biol. Chem. 267, 3932.
Toeplitz, B.K., et al. 1979. J. Am. Chem. Soc.101, 3344.
Liu, C.M., and Hermann, T.E. 1978. J. Biol. Chem.253, 5892.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaël Boily et al.
Journal of the American Heart Association, 10(17), e020608-e020608 (2021-08-31)
Background Angiotensin II (Ang II), a critical mediator of hypertension, impairs neurovascular coupling. Since astrocytes are key regulators of neurovascular coupling, we sought to investigate whether Ang II impairs neurovascular coupling through modulation of astrocytic Ca2+ signaling. Methods and Results
Gi-Cheon Kim et al.
Frontiers in oncology, 10, 642-642 (2020-06-02)
ETS1 has shown dichotomous roles as an oncogene and a tumor suppressor gene in diverse cancers, but its functionality in breast cancer tumorigenesis still remains unclear. We utilized the Cancer Genome Atlas (TCGA) database to analyze comprehensive functions of ETS1
Thijs Roebroek et al.
Nature communications, 12(1), 2005-2005 (2021-04-02)
Förster resonant energy transfer (FRET) is a powerful mechanism to probe associations in situ. Simultaneously performing more than one FRET measurement can be challenging due to the spectral bandwidth required for the donor and acceptor fluorophores. We present an approach
Lynn Sanford et al.
ACS chemical neuroscience, 11(4), 508-514 (2020-02-06)
Ionic Zn2+ has increasingly been recognized as an important neurotransmitter and signaling ion in glutamatergic neuron pathways. Intracellular Zn2+ transiently increases as a result of neuronal excitation, and this Zn2+ signal is essential for neuron plasticity, but the source and
Xiaozheng Xu et al.
The Journal of cell biology, 219(6) (2020-05-22)
Blockade antibodies of the immunoinhibitory receptor PD-1 can stimulate the anti-tumor activity of T cells, but clinical benefit is limited to a fraction of patients. Evidence suggests that BTLA, a receptor structurally related to PD-1, may contribute to resistance to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service